Skip to Content
Merck
All Photos(1)

Documents

MABE71

Sigma-Aldrich

Anti-Histone Antibody, Pan, clone F152.C25.WJJ

clone F152C25W, from mouse

Synonym(s):

Histone H1.4, Histone H1b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

F152C25W, monoclonal

species reactivity

human, bovine

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H2AC21(317772)

General description

This antibody detects histone H1 and nuclesomal core proteins histones H2a, H2b, H3, H4. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Histone H1 is a nuclear protein necessary for condensation of nucleosome chains into higher order structures.

Application

Anti-Histone Antibody, Pan, clone F152.C25.WJJ is a Mouse Monoclonal Antibody for detection of Histone also known as Histone H1.4, Histone H1b & has been validated in WB, ELISA.
ELISA Analysis: A representative lot was used by an independent laboratory in ELISA. (Mizoguchi, E., et al. (1997). Gastroenterology. 113:1828-1835.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blot in Jurkat cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Histone H1 and core proteins on 10 µg of Jurkat cell lysate.

Target description

~14-33 kDa observed

Linkage

Replaces: MAB052

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeynep Eren-Ghiani et al.
Cell reports, 13(11), 2327-2335 (2015-12-18)
The formation of motile spermatozoa involves the highly conserved formation of protamine-rich, tightly packed chromatin. However, genetic loss of protamine function in Drosophila and mice does not lead to significant decompaction of sperm chromatin. This indicates that other proteins act
Drosophila protamine-like Mst35Ba and Mst35Bb are required for proper sperm nuclear morphology but are dispensable for male fertility.
Tirmarche, S; Kimura, S; Sapey-Triomphe, L; Sullivan, W; Landmann, F; Loppin, B
G3 (Bethesda, Md.) null
Vanessa de Carvalho Oliveira et al.
Frontiers in immunology, 14, 1042686-1042686 (2023-02-11)
Neutrophil extracellular traps (NETs) serve to immobilize and kill pathogens, but also can contribute to the progression of several inflammatory and auto-immune diseases, as well as cancer. Whence the importance of elucidating the mechanisms underlying NET formation. In this regard
Daniela Torres-Campana et al.
PLoS genetics, 16(3), e1008543-e1008543 (2020-03-07)
Following fertilization of a mature oocyte, the formation of a diploid zygote involves a series of coordinated cellular events that ends with the first embryonic mitosis. In animals, this complex developmental transition is almost entirely controlled by maternal gene products.
Louise D McCullough et al.
The Journal of clinical investigation, 131(17) (2021-09-29)
Inter-α inhibitor proteins (IAIPs) are a family of endogenous plasma and extracellular matrix molecules. IAIPs suppress proinflammatory cytokines, limit excess complement activation, and bind extracellular histones to form IAIP-histone complexes, leading to neutralization of histone-associated cytotoxicity in models of sepsis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service