Skip to Content

MABE175

Anti-PML Isoform II Antibody, clone 1A8.1

clone 1A8.1, from mouse

Synonym(s):

PML-2, PML-II, Protein PML isoform II, Promyelocytic leukemia protein isoform II, RING finger protein 71 isoform II, TRIM19kappa, Tripartite motif-containing protein 19 isoform II

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1A8.1, monoclonal
Application:
ICC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A8.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PML(5371)

General description

Protein PML (UniProt P29590; also known as Promyelocytic leukemia protein, RING finger protein 71, Tripartite motif-containing protein 19) is encoded by the PML (also known as MYL, PP8675, RNF71, TRIM19) gene (Gene ID 5371) in human. The human PML gene/protein adopted its name when it was originally discovered as an RARα fusion protein caused by chromosomal translocation in acute promyelocytic leukemia (APL) patients. In normal cells, the PML protein interacts with multiple protein partners in the nucleus to form PML nuclear bodies (PML-NBs) that mediate a variety of cellular functions including senescence, apoptosis, protein degradation, telemere stasbility, and antiviral defense. At least 12 PML isoforms are generated by alternative splicings, including six nuclear isoforms (PML-I to PML-VI) and the cytoplasmic isoform PML-VII (a.k.a. PML-VIIB), other spliced isoforms without exon 6-coding sequence (a.a. 467-552) are likewise expected to be cytoplasmic due to the abscence of nuclear localization signal (NLS) encoded by exon 6. All isoforms share the same N-terminal region (a.a. 1-419) containing the RBCC/tripartite motif and may have related functions due to their common functional RBCC/TRIM domain, while increasing evidences also suggest that each PML isoform possesses distinct functions mediated by its specific C-terminal sequence. Isoform PML-1 (a.k.a. PML-I, TRIM19alpha; UniProt P29590-1) and PML-2 (a.k.a. PML-II or TRIM19kappa; UniProt P29590-8) share the same N-terminal 570-a.a. sequence (encoded by exons 1 to 7a), however PML-1 derives its remaining C-terminal sequence (a.a. 571-882) from exons 8a & 9, while PML-II derives its (a.a. 571-829) from exon 7b with a different reading frame as that of PML-V.
~110 kDa observed. 90.72 kDa calculated. Uncharacterized band(s) may appear in some lysates.

Immunogen

Epitope: C-terminal region.
KLH-conjugated linear peptide corresponding to a C-terminal sequence of human PML Isoform II.

Application

Anti-PML Isoform II Antibody, clone 1A8.1 is an antibody against PML Isoform II for use in Western Blotting, Immunocytochemistry.
Immunocytochemistry Analysis: 40 µg/mL from a representative lot immunostained 10% neutral-buffered formalin-fixed HEK293 cells transfected with human PML isoform II by non-fluorescent immunocytochemistry (Courtesy of Professor Ygal Haupt, Peter MacCallum Cancer Centre, East Melbourne, Australia).
Immunocytochemistry Analysis: 10 µg/mL from a representative lot immunostained 4% paraformaldehyde-fixed HEK293 cells transfected with human PML isoform II by fluorescent immunocytochemistry (Courtesy of Professor Ygal Haupt, Peter MacCallum Cancer Centre, East Melbourne, Australia).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Biochem/physiol Actions

Clone 1A8.1 was raised against an C-terminal region sequence unique to PML isoform II (PML-2), but not present in the other 11 PML spliced isoforms reported by UniProt (P29590).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in lysate from PML-2 transfected HEK293 cells.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected the exogenously expressed human PML-2 in 10 µg of lysate from transfected HEK293 cells.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library