Skip to Content

MABC640

Anti-MUM1 Antibody, clone 4G10

ascites fluid, clone 4G10, from mouse

Synonym(s):

PWWP domain-containing protein MUM1, Mutated melanoma-associated antigen 1, MUM-1, Protein expandere

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
4G10, monoclonal
Application:
IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4G10, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PWWP3A(84939)

General description

Mutated melanoma-associated antigen 1 (MUM1), also known as Protein expandere and PWWP domain-containing protein MUM1. MUM1’s normal role is as a DNA and chromatin associated protein that is involved in the cell’s response to DNA damage. It is driven to DNA breaks by its interaction with another DNA chromatin protein, TP53BP1, which it binds. The binding facilitates chromatin relaxation and entry of DNA repair enzymes. MUM1 is required for efficient DNA repair and cellular survival after DNA damage. Interestingly MUM1 was first identified by cloning the chromosomal breakpoints of a multiple myeloma cell line and was identified as a member of the interferon response transcriptional factor gene family (IFR). MUM1 appears to play a critical role in cell proliferation, differentiation and survival of various cancers, as well as monocytes and dendritic cells. MUM1 is expressed in B cells and plasma cells as well and in some populations of T lymphocytes. EMD-Millipore’s Anti-MUM1 mouse monoclonal antibody has been tested in against recombinantly expressed human MUM1 protein and in paraffin embedded immunohistochemistry on human melanoma, non-Hodgkin’s lymphoma and normal uterus tissue sections.
~55 kDa observed

Immunogen

Purified recombinant fragment of human MUM1 expressed in E. Coli.

Application

Detect Mutated melanoma-associated antigen 1 using this mouse monoclonal antibody, Anti-MUM1 Antibody, clone 4G10 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected MUM1 in human uterus, melanoma, and non-Hodgkin`s lymphoma tissues.

Optimal working dilutions must be determined by end user.

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Recombinant MUM1 protein
Evaluated by Western Blotting in recombinant MUM1 protein.

Western Blotting Analysis: A 1:500-2000 dilution of this antibody detected recombinant MUM1.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library