Skip to Content
Merck
All Photos(1)

Key Documents

MABC1236

Sigma-Aldrich

Anti-DNA-PKcs Antibody, clone 3A9.1

clone 3A9.1, from mouse

Synonym(s):

DNA-dependent protein kinase catalytic subunit, DNA-PK catalytic subunit, DNA-PKcs, DNPK1, p460

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
clone:
3A9.1, monoclonal
application:
WB
species reactivity:
human
technique(s):
western blot: suitable
citations:
3

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A9.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PRKDC(5591)

General description

DNA-dependent protein kinase catalytic subunit (EC 2.7.11.1; UniProt P78527; also known as DNA-PK catalytic subunit, DNA-PKcs, DNPK1, p460) is encoded by the PRKDC (also known as HYRC, HYRC1, IMD26, XRCC7) gene (Gene ID 5591) in human. DNA-PK is a heterotrimer of PRKDC and the Ku p70/YRCC6-p86/XRCC5 dimer. It acts as a molecular sensor for DNA damage and is involved in DNA non-homologous end joining required for double-strand break (DSB) repair and V(D)J recombination. Upon binding to DNA, it expresses its catalytic properties. DNA-PKcs falls into the phosphatidylinositol (PI) 3-kinase family, however, biochemical assays indicate that it phosphorylates proteins and has no detectable activity toward lipids. DNA-PKcs is displays high similarity to PI kinase family members involved in cell cycle control, DNA repair, and DNA damage responses. Deficiency of DNA-PKcs is linked to severe combined immunodeficiency characterized by reduced or absent T and B cells, recurrent candidiasis, and lower respiratory tract infections. Ref.: Hartley, KO., et al. (1995). 82, 849-856.

Specificity

Clone 3A9.1 detects DNA-PK catalytic subunit (DNA-PKcs), but not the regulatory subunit (Ku). Target epitope is present in both spliced isoforms of human DNA-PKcs reported by UniProt (P78527).

Immunogen

GST-tagged recombinant fragment from the region between FAT and KIP-binding domain of human DNA-PKcs

Application

Detect DNA-PK catalytic subunit (DNA-PKcs) using this mouse monoclonal Anti-DNA-PKcs Antibody, clone 3A9.1, Cat. No. MABC1236, validated for use in Western Blotting.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected DNA-PKcs in 10 µg of K562 cell lysate.

Target description

~470 kDa observed. 469.1/466.5 kDa (isoform 1/2) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Roulston et al.
Molecular cancer therapeutics, 21(2), 245-256 (2021-12-17)
Ataxia telangiectasia and Rad3-related (ATR) kinase protects genome integrity during DNA replication. RP-3500 is a novel, orally bioavailable clinical-stage ATR kinase inhibitor (NCT04497116). RP-3500 is highly potent with IC50 values of 1.0 and 0.33 nmol/L in biochemical and cell-based assays
Mzwanele Ngubo et al.
Aging cell, 23(7), e14150-e14150 (2024-04-05)
Hutchinson-Gilford Progeria syndrome (HGPS) is a lethal premature aging disorder caused by a de novo heterozygous mutation that leads to the accumulation of a splicing isoform of Lamin A termed progerin. Progerin expression deregulates the organization of the nuclear lamina
N V Srikanth Vallabani et al.
Mutagenesis, 34(3), 265-277 (2019-07-05)
Zinc oxide nanoparticles (ZnO NPs) with their wide range of consumer applications in day-to-day life received great attention to evaluate their effects in humans. This study has been attempted to elucidate the DNA damage response mechanism in a dermal model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service