Skip to Content
Merck
All Photos(1)

Key Documents

MAB925

Sigma-Aldrich

Anti-Rubella Antibody, E1, clone EI-20

clone EI-20, Chemicon®, from mouse

Synonym(s):

German Measles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

EI-20, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Specific for E1. The antibody recognizes an epitope located between amino acids 202 and 283 of the E1 glycoprotein.

Immunogen

Epitope: E1

Application

Anti-Rubella Antibody, E1, clone EI-20 is an antibody against Rubella for use in IF.
IFA.

Western blot

Neutralizes infectivity.

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Purified immunoglobulin presented as a liquid in 0.2 M phosphate buffer, 0.25 M NaCl with 0.1% sodium azide.

Storage and Stability

Maintain at +2-8°C for up to 6 months in undiluted aliquots.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliane Schulz et al.
International journal of molecular sciences, 24(9) (2023-05-13)
Vertical transmission of rubella virus (RuV) occurs at a high rate during the first trimester of pregnancy. The modes of vertical transmission including the response of trophoblasts to RuV are not well understood. Here, RuV-trophoblast interaction was studied in the
Diagnosis of recent primary rubella virus infections: Significance of glycoprotein-based IgM serology, IgG avidity and immunoblot analysis.
Wandinger KP, Saschenbrecker S, Steinhagen K, Scheper T, Meyer W, Bartelt U, Enders G
Journal of Virological Methods null
Nicole C Bilz et al.
Journal of virology, 92(17) (2018-06-29)
The flexible regulation of cellular metabolic pathways enables cellular adaptation to changes in energy demand under conditions of stress such as posed by a virus infection. To analyze such an impact on cellular metabolism, rubella virus (RV) was used in
Monoclonal antibody-defined epitope map of expressed rubella virus protein domains
Wolinsky, J. et al.
Journal of virology, 65, 3986-3994 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service