Skip to Content
Merck
All Photos(1)

Documents

MAB8887

Sigma-Aldrich

Anti-Collagen Type II Antibody, clone 6B3

clone 6B3, Chemicon®, from mouse

Synonym(s):

Anti-Anti-ANFH, Anti-Anti-AOM, Anti-Anti-COL11A3, Anti-Anti-SEDC, Anti-Anti-STL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6B3, monoclonal

species reactivity

human, chicken, mouse, salamander

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... COL2A1(1280)

Specificity

In Western blotting (but not ELISA), MAB8887 recognizes both a1(II) and a3(XI) chains which have identical primary structure. In Western blotting, MAB8887 reacts with the TCA fragment of lathyritic type II collagen after digestion with mammalian collagenase. It also reacts with pepsin-digested type II collagen. It will not react with intact Collagen II under traditional conditions.

Its epitope is localized in the triple helix of type II collagen. It shows no cross-reaction with type I or type III collagen. Immunoblotting of CNBr peptides of collagen II shows that MAB8887 reacts with CB11 (25kDa) fragment which is the site of immunogenic and arthritogenic epitopes along the intact type II molecule.

In pepsin solublized collagen II, MAB8887 reacts with a 95-97 kDa fragment, as well as, the native single chain of 120kDa. If propeptides are present, MAB8887 will detect a 200kDa fragment.

Immunogen

Purified type II collagen; obtained from adult chicken sternum by limited pepsinization.

Application

Anti-Collagen Type II Antibody, clone 6B3 is an antibody against Collagen Type II for use in IF, IH & WB.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
Western Blot (Mayne, 1994)

Immunohistochemistry (Formalin/Paraffin): 1-2 μg/mL, 30 minutes at room temperature. Staining of paraffin embedded tissues requires digestion of tissue sections with pepsin at 1mg/mL in Tris HCl, pH 2.0 for 15 minutes at RT or 10 minutes at 37ºC.

Immunofluorescence

ELISA

Optimal working dilutions must be determined by end user.

Target description

120 kDa

Physical form

Format: Purified
Protein G Purified
Purified immunoglobulin in 10 mM PBS, pH 7.4 with 0.2% BSA and 0.09% sodium azide.

Storage and Stability

Maintain at 2–8°C in undiluted aliquots for up to 12 months

Analysis Note

Control
Cartilage in lung or fetus

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Qiang Fu et al.
Molecular medicine reports, 13(6), 4659-4665 (2016-04-16)
The aim of the present study was to evaluate the tumorigenic potential of nuclear transfer-derived (nt) mouse embryonic stem cells (mESCs) transplanted into infarcted rat hearts. The nt‑mESCs were cultured using a bioreactor system to develop embryoid bodies, which were
Jungsun Lee et al.
Cell transplantation, 26(10), 1673-1687 (2017-12-19)
Given recent progress in regenerative medicine, we need a means to expand chondrocytes in quantity without losing their regenerative capability. Although many reports have shown that growth factor supplementation can have beneficial effects, the use of growth factor-supplemented basal media
Prostaglandin E2 (PGE2) exerts biphasic effects on human tendon stem cells.
Zhang, J; Wang, JH
Testing null
R S Cosden et al.
Osteoarthritis and cartilage, 19(2), 200-205 (2010-12-01)
The ability to fully regenerate lost limbs has made the axolotl salamander (Ambystoma mexicanum) a valuable model for studies of tissue regeneration. The current experiments investigate the ability of these vertebrates to repair large articular cartilage defects and restore normal
Low-density lipoprotein receptor-related protein 5 governs Wnt-mediated osteoarthritic cartilage destruction.
Shin, Y; Huh, YH; Kim, K; Kim, S; Park, KH; Koh, JT; Chun, JS; Ryu, JH
Arthritis Research & Therapy null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service