MAB821
Anti-Herpes Simplex Virus I & II Antibody, clone 4F10.3
clone 4F10.3, Chemicon®, from mouse
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
clone
4F10.3, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
immunofluorescence: suitable
isotype
IgG2a
shipped in
wet ice
Specificity
Checked by immunoenzymatic method and shown to be highly reactive to HSV-I strains "Mac" and "F", moderately reactive to HSV-II strain "G" and weakly reactive to HSV-II strain "Ms". Possible application: direct detection of HSV I and HSV II virus.
Application
Detect Herpes Simplex Virus I & II using this Anti-Herpes Simplex Virus I & II Antibody, clone 4F10.3 validated for use in ELISA & IF.
ELISA Type I 1:6400 Type II 1:100
IFA infected fibroblasts 1:800
Final working dilutions must be determined by end user.
IFA infected fibroblasts 1:800
Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Infectious Diseases - Viral
Physical form
Format: Purified
Purified immunoglubulin. Liquid in 0.02M PB with 0.25M NaCl.pH=7.6, 0.1% Sodium Azide.
Storage and Stability
Maintain at 2-8°C.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J M McSharry et al.
Journal of clinical microbiology, 36(4), 958-964 (1998-05-23)
A flow cytometric assay has been developed for the measurement of susceptibilities to ganciclovir of laboratory strains and clinical isolates of human cytomegalovirus (HCMV). The assay uses fluorochrome-labeled monoclonal antibodies to HCMV immediate-early and late antigens to identify HCMV-infected cells
Jianyun Liu et al.
Immunology, 140(2), 191-201 (2013-05-29)
CD1d-restricted T (natural killer T; NKT) cells are important for controlling herpesvirus infections. Interestingly, herpes simplex virus (HSV) can down-regulate CD1d-mediated activation of NKT cells. We have previously shown that the Thr322 residue in the cytoplasmic tail of human CD1d
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service