Skip to Content
Merck
All Photos(1)

Key Documents

MAB819

Sigma-Aldrich

Anti-Parainfluenza 1, 2, 3 Blend Antibody, clones 251-12A, 354-8B, 343-5G, 231-9F, 233-4D, 341-7H

ascites fluid, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

231-9F, monoclonal
233-4D, monoclonal
251-12A, monoclonal
341-7H, monoclonal
343-5G, monoclonal
354-8B, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable

isotype

IgG

shipped in

wet ice

Specificity

Blend of six monoclonal antibodies with specificities for FO/F1, NP and HN proteins. Reacts with all known serotypes of Parainfluenza I, II and III.

Immunogen

A blend of clones separately immunized with C-35, Greer, and C2 43.

Application

ELISA at 1:800+.

Indirect Immunofluorescence at 1:100 -200+ (fresh frozen tissue sections).

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Parainfluenza 1, 2, 3 Blend Antibody, clones 251-12A, 354-8B, 343-5G, 231-9F, 233-4D, 341-7H is validated for use in ELISA, IF for the detection of Parainfluenza 1.

Physical form

Ascites fluid with 0.1% sodium azide as a preservative.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Parainfluenza 1, 2, 3 Control Slides, Catalogue Number 5011-5

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pathology of parainfluenza virus infection in patients with congenital immunodeficiency syndromes.
John F Madden, James L Burchette, Laura P Hale
Human Pathology null
Human parainfluenza virus type 3 in wild nonhuman primates, Zambia.
Sasaki, M; Ishii, A; Orba, Y; Thomas, Y; Hang'ombe, BM; Moonga, L; Mweene, AS; Ogawa et al.
Emerging Infectious Diseases null
J S Ellis et al.
Journal of clinical microbiology, 35(8), 2076-2082 (1997-08-01)
Multiple-target (multiplex) reverse transcription-PCR (RT-PCR) for detection, typing, and subtyping of the hemagglutinin gene of influenza type A (H3N2 and H1N1) and type B viruses was developed and applied prospectively to virological surveillance of influenza in England in the 1995-1996

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service