Skip to Content
Merck
All Photos(1)

Key Documents

MAB342A4

Sigma-Aldrich

Anti-Galactocerebroside Antibody, clone mGalC, Alexa Fluor488 Conjugate

clone mGalC, from mouse, ALEXA FLUOR 488

Synonym(s):

Galactoceramide, Galactocerebroside, Galactosylceramide, GalC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

mGalC, monoclonal

species reactivity

rabbit, rat

species reactivity (predicted by homology)

human, bovine (immunogen homology), chicken, mouse

technique(s)

immunocytochemistry: suitable

isotype

IgG3

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GALC(2581)

General description

Galactocerebroside (GalC) is a major galactosphingolipid of myelin which plays a role in myelination. GalC is a very useful, specific marker for oligodendroglial lineage.

Specificity

This antibody recognizes Galactocerebroside (GalC), sulfatide, psychosine and other galactolipids. Cross-reacts with the sulfatide ester of GalC, but to a 16-fold to a lesser extent. No cross-reactivity with sphingosine, ceramide, mixed ganglioside or glucocerebroside. Binds specifically with oligodendrocytes and Schwann cells.

Application

Detect Galactocerebroside using this Anti-Galactocerebroside Antibody, clone mGalC, Alexa Fluor488 Conjugate validated for use in ICC.

Quality

Evaluated by Immunocytochemistry in rat oligodendrocyte cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Galactocerebroside in rat oligodendrocyte cells. Performed without Triton.

Physical form

Purified mouse monoclonal IgG3 conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Analysis Note

Control
Rat oligodendrocyte cells

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shun Yamamuro et al.
International journal of oncology, 47(5), 1647-1654 (2015-09-24)
Glioma stem-like cells (GSCs) are undifferentiated cells that are considered to be an origin of glioblastomas. Furthermore, they may contribute to treatment resistance and recurrence in glioblastomas. GSCs differentiate into differentiated glioma cells (non-glioma stem-like cells: non‑GSCs), and interconversion might
Juan F Reyes et al.
Glia, 62(3), 387-398 (2014-01-03)
The origin of α-synuclein (α-syn)-positive glial cytoplasmic inclusions found in oligodendrocytes in multiple system atrophy (MSA) is enigmatic, given the fact that oligodendrocytes do not express α-syn mRNA. Recently, neuron-to-neuron transfer of α-syn was suggested to contribute to the pathogenesis
Hal X Nguyen et al.
The Journal of comparative neurology, 522(12), 2767-2783 (2014-04-10)
Human embryonic stem cells (hESC) and induced pluripotent stem cells (hiPSC) can differentiate into many cell types and are important for regenerative medicine; however, further work is needed to reliably differentiate hESC and hiPSC into neural-restricted multipotent derivatives or specialized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service