Skip to Content
Merck
All Photos(5)

Documents

MAB302

Sigma-Aldrich

Anti-Glutamine Synthetase Antibody, clone GS-6

clone GS-6, Chemicon®, from mouse

Synonym(s):

Anti-Glutamine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

GS-6, monoclonal

species reactivity

human, rat, mouse, sheep

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GLUL(2752)

General description

Glutamine Synthetase catalyzes the conversion of ammonia and glutamate to glutamine. It is found in astrocytes as an octamer of identical 45 kDa subunits. The function of Glutamine Synthetase is the detoxification of brain ammonia. It also has an important role in the metabolic regulation of neurotransmitter glutamate. Because of the multiple functions and importance of Glutamine Synthetase in cellular metabolism, both catalytic activities and synthesis are highly regulated. The activity of Glutamine Synthetase is controlled by adenylylation. Its activity is decreased in the cerebral cortex of brains affected by Alzheimer′s disease, particularly in the vicinity of senile plaques. It is also decreased under conditions of glucose deprivation. The level of expression of Glutamine Synthetase is increased during ischemia in vivo or hypoxia in culture.

Specificity

Reacts strongly with a single band on Western blots of sheep brain and rat brain. Glutamine synthetase, an enzyme that catalyzes the amination of glutamic acid to form glutamine, is found in mammals as an octamer of eight identical 45 kDa subunits. Glutamine synthetase activity has been shown to be a useful marker of astrocytes and an important differentiation feature in retina. Glutamine synthetase is also considered to be a key enzyme in the recycling of the neurotransmitter glutamate.

Immunogen

Glutamine synthetase purified from sheep brain.

Application

Detect Glutamine Synthetase using this Anti-Glutamine Synthetase Antibody, clone GS-6 validated for use in ELISA, IH, IH(P) & WB with more than 45 product citations.
Research Category
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism
Western blot: 1:1,000-1:5,000

Immunohistochemistry on rat brain

ELISA

Optimal working dilutions must be determined by end user.

Target description

45 kDa

Physical form

Format: Purified
Protein A purified
Purified immunoglobulin. Liquid in buffer containing 50% glycerol, BSA and 0.09% sodium azide.

Storage and Stability

Stable for 1 year at -20ºC in undiluted aliquots from date of receipt.

Analysis Note

Control
Rat brain tissue, rat brain cytosolic fraction extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alterations in protein expression and membrane properties during Muller cell gliosis in a murine model of transient retinal ischemia.
Hirrlinger PG, Ulbricht E, Iandiev I, Reichenbach A, Pannicke T
Neuroscience Letters null
H Ueda et al.
Investigative ophthalmology & visual science, 41(12), 3908-3914 (2000-10-29)
Dystrobrevin is a newly discovered dystrophin-associated protein with multiple sites for phosphorylation on tyrosine residues. In the present study, the cellular distribution and subcellular localization of dystrobrevin were examined in the adult rat retina, cornea, lens, iris, ciliary body, and
Markel Olabarria et al.
Molecular neurodegeneration, 6, 55-55 (2011-08-02)
Astrocytes are fundamental for brain homeostasis and the progression and outcome of many neuropathologies including Alzheimer's disease (AD). In the triple transgenic mouse model of AD (3xTg-AD) generalised hippocampal astroglia atrophy precedes a restricted and specific β-amyloid (Aβ) plaque-related astrogliosis.
Effects of retinal glial cells on isolated rat retinal ganglion cells.
K Kashiwagi, Y Iizuka, M Araie, Y Suzuki, S Tsukahara
Investigative Ophthalmology & Visual Science null
Redistribution of astrocytic glutamine synthetase in the hippocampus of chronic epileptic rats.
Papageorgiou IE, Gabriel S, Fetani AF, Kann O, Heinemann U.
Glia null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service