Skip to Content
Merck
All Photos(1)

Key Documents

MAB2030

Sigma-Aldrich

Anti-Chondroitin 4 Sulfate Antibody, clone BE-123

ascites fluid, clone BE-123, Chemicon®

Synonym(s):

Anti-CSPG4A, Anti-HMW-MAA, Anti-MCSP, Anti-MCSPG, Anti-MEL-CSPG, Anti-MSK16, Anti-NG2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

BE-123, monoclonal

species reactivity

human, canine, rat, sheep, bovine, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CSPG4(1464)

Specificity

Reacts with both mouse (fetal and adult) and human (fetal and adult) chondroitinase ABC digested cartilage proteoglycans. Chondroitin-4 sulfate disaccharides successfully inhibit the binding of monoclonal antibody to proteoglycans. No reaction with native proteoglycans or with proteoglycans digested with either chondroitinase AC or testicular hyaluronidase.

Immunogen

Mouse proteoglycan.

Application

Anti-Chondroitin 4 Sulfate Antibody, clone BE-123 detects level of Chondroitin 4 Sulfate & has been published & validated for use in ELISA, RIA & WB.
ELISA at 1:400-1:3,200.

RIA at 1:50-1:6,400.

Immunohistochemistry: This antibody only reacts with digested material. Fixed tissue must be digested with chondroitinase ABC (Sigma).
The Reference 23757342 should be indicated for IHC. (Reine, TM., et al., PubMed ID: 23757342)

Western Blot: 1:1000-1:4000 using ECL detection method. Chondroitinase ABC treatment of lysate sample is helpful.
In any alkali buffer, e.g. Tris or acetate (50mM or 100mM) pH 7.4-7.6. Make a stock solution by diluting one vial of Sigma Chondroitinase A, B, C, (5 units/vial) in 100 µL . For use, dilute stock 10-fold into the buffer of choice. Use at a ratio of 50-100mg of proteoglycan per 5 units of enzyme. On fixed cells, dilute in Tris acetate and on non-fixed cellsy dilute 10-25 fold in DMEM.
Poole, C.A., et al., J. Histochem. Cytochem. 1991. 39:1175-1187.

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Ascites fluid containing no preservatives.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Schnabel et al.
Osteoarthritis and cartilage, 10(1), 62-70 (2002-02-14)
The aim of the present study was the investigation of differential gene expression in primary human articular chondrocytes (HACs) and in cultivated cells derived from HACs. Primary human articular chondrocytes (HACs) isolated from non-arthritic human articular cartilage and monolayer cultures
Myriam Baratin et al.
PloS one, 2(2), e228-e228 (2007-02-22)
Host innate immunity contributes to malaria clinical outcome by providing protective inflammatory cytokines such as interferon-gamma, and by shaping the adaptive immune response. Plasmodium falciparum (Pf) is the etiologic agent of the most severe forms of human malaria. Natural Killer
Karen W Sanden et al.
Journal of biophotonics, 12(9), e201800436-e201800436 (2019-06-05)
In the present study, Fourier-transform infrared spectroscopy (FTIR) is investigated as a method to measure connective tissue components that are important for the quality of Atlantic cod filets (Gadus morhua L.). The Atlantic cod used in this study originated from
E Ortega-Barria et al.
The Journal of biological chemistry, 274(3), 1267-1276 (1999-01-09)
Toxoplasma gondii is one of the most widespread parasites of humans and animals. The parasite has a remarkable ability to invade a broad range of cells within its mammalian hosts by mechanisms that are poorly understood at the molecular level.
Chelsea S Sullivan et al.
Scientific reports, 8(1), 6143-6143 (2018-04-20)
Perineuronal nets (PNNs) are implicated in closure of critical periods of synaptic plasticity in the brain, but the molecular mechanisms by which PNNs regulate synapse development are obscure. A receptor complex of NCAM and EphA3 mediates postnatal remodeling of inhibitory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service