Skip to Content
Merck
All Photos(1)

Documents

MAB1676

Sigma-Aldrich

Anti-Talin 1 Antibody, NT, a.a. 139-433, clone TA205

clone TA205, Chemicon®, from mouse

Synonym(s):

Anti-ILWEQ, Anti-TLN, Anti-talin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

TA205, monoclonal

species reactivity

rabbit, human

should not react with

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TLN1(7094)

Specificity

Human Talin. This antibody reacts with an N-terminal epitope in talin, between amino acids 139-433. On microinjection into whole cells MAB1676 disrupts actin stress fibers and focal contacts and inhibits cell motility.

Immunogen

Epitope: N-terminus, a.a. 139-433

Application

Anti-Talin 1 Antibody, N-terminus, a.a. 139-433, clone TA205 detects level of Talin & has been published & validated for use in IP, WB, IH.
Immunoblotting, recognizes a band at ~230kDa.

Immunoprecipitation

Immunofluorescence

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Liquid in 0.02 M PB pH 7.6, 0.25 M NaCl containing 0.1% sodium azide.

Storage and Stability

Maintain between 2 and 8°C for up to six months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen P Fong et al.
Cellular microbiology, 8(11), 1753-1767 (2006-07-11)
Actinobacillus actinomycetemcomitans produces a leukotoxin (Ltx) that kills leukocyte function-associated antigen-1 (LFA-1)-bearing cells from man, the Great Apes and Old World monkeys. The unique specificity of Ltx for the beta2 integrin, LFA-1, suggests it is capable of providing insight into
Masaru Niki et al.
Thrombosis and haemostasis, 115(5), 969-978 (2016-01-23)
Adaptor proteins play a critical role in the assembly of signalling complexes after engagement of platelet receptors by agonists such as collagen, ADP and thrombin. Recently, using proteomics, the Dok (downstream of tyrosine kinase) adapter proteins were identified in human
Karen P Fong et al.
Blood advances, 5(23), 4901-4909 (2021-09-28)
Blood clot contraction is driven by traction forces generated by the platelet cytoskeleton that are transmitted to fibrin fibers via the integrin αIIbβ3. Here we show that clot contraction is impaired by inhibitors of the platelet cytosolic protease calpain. We
Maria Salmela et al.
Scientific reports, 7(1), 3414-3414 (2017-06-15)
Conformational activation of integrins is generally required for ligand binding and cellular signalling. However, we have previously reported that the nonactivated conformation of α2β1 integrin can also bind to large ligands, such as human echovirus 1. In this study, we
Daniel Friedman et al.
Journal of cell science, 134(7) (2021-03-14)
Natural killer (NK) cells can kill infected or transformed cells via a lytic immune synapse. Diseased cells may exhibit altered mechanical properties but how this impacts NK cell responsiveness is unknown. We report that human NK cells were stimulated more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service