Skip to Content
Merck
All Photos(1)

Documents

MAB1358

Sigma-Aldrich

Anti-Integrin α6B Antibody, cytoplasmic domain, clone 6B4

clone 6B4, Chemicon®, from mouse

Synonym(s):

CD49f

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6B4, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA6(3655)

General description

Integrins are a family of heterodimeric membrane glycoproteins consisting of non-covalently associated alpha and beta subunits. More than 18 alpha and 8 beta subunits with numerous splice variant isoforms have been identified in mammals. In general, integrin function as receptors for extracellular matrix proteins. Certain integrins can also bind to soluble ligands or to counter-receptors on adjacent cells, such as the intracellular adhesion molecules (ICAMs), resulting in aggregation of cells. Signals transduced by integrins play a role in many biological processes, including cell growth, differentiation, migration and apoptosis. For integrin subunits alpha 3 and alpha6, two cytoplasmic variants, A and B, have been identified.

Specificity

MAB1358 recognizes specifically the cytoplasmic domain of integrin subunit alpha 6B. The antibody stains the basement membrane zone of ducts and acini of salivary gland, kidney tubules, epithelium of colonic crypts and weakly stains sweat glands and sebaceous glands.

Broad species reactivity is expected due to the conserved nature of the epitope.

Immunogen

Synthetic Peptide to the cytoplasmic domain of integrin subunit alpha6B including an appending N-terminal cysteine: CSRYDDSVPRYHAVRIRKEEREIKDEKYIDNLEKK

Application

Anti-Integrin α6B Antibody, cytoplasmic domain, clone 6B4 is an antibody against Integrin α6B for use in WB, IC, IH.
Western blotting: 1:100-1:500

Immunocytochemistry: 1:100-1:500 on frozen sections, 1:50-1:100 with avidin-biotin complex detection

Immunocytochemistry

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified

Storage and Stability

Store at 2-8°C, or -20°C in small aliquots for up to 12 months after date of receipt. Avoid freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darcie D Seachrist et al.
The Journal of biological chemistry, 295(33), 11707-11719 (2020-06-25)
The phenotypes of each breast cancer subtype are defined by their transcriptomes. However, the transcription factors that regulate differential patterns of gene expression that contribute to specific disease outcomes are not well understood. Here, using gene silencing and overexpression approaches
Dimpi Mukhopadhyay et al.
Cell reports, 42(11), 113347-113347 (2023-11-01)
Understanding the cell biological mechanisms that enable tumor cells to persist after therapy is necessary to improve the treatment of recurrent disease. Here, we demonstrate that transient receptor potential channel 6 (TRPC6), a channel that mediates calcium entry, contributes to
Reconstruction of a human hemicornea through natural scaffolds compatible with the growth of corneal epithelial stem cells and stromal keratocytes.
Barbaro, V; Ferrari, S; Fasolo, A; Ponzin, D; Di Iorio, E
Molecular Vision null
Seo-Hyun Choi et al.
Molecular cancer research : MCR, 20(8), 1305-1319 (2022-04-09)
KRAS mutation in colorectal cancer is associated with aggressive tumor behavior through increased invasiveness and higher rates of lung metastases, but the biological mechanisms behind these features are not fully understood. In this study, we show that KRAS-mutant colorectal cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service