LSKMAGHDKIT
PureProteome Human Albumin/Immunoglobulin Depletion Kit
The PureProteome Human Albumin/Immunoglobulin Depletion Kit is a magnetic bead based kit that enables high depletion efficiency of Albumin and all Immunoglobulins from human serum or plasma samples.
Synonym(s):
Protein Purification
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116133
eCl@ss:
32160405
NACRES:
NA.56
Recommended Products
packaging
kit of 12 mL beads
manufacturer/tradename
PureProteome
technique(s)
depletion: suitable (human serum/plasma)
protein purification: suitable
particle size
10 μm
shipped in
wet ice
storage temp.
2-8°C
General description
The PureProteome Human Albumin/Immunoglobulin Depletion Kit is a magnetic bead based kit that enables high depletion efficiency (typically >99%) of Albumin and all Immunoglobulins (i.e IgG, IgA, IgM, IgE and IgD) from human serum or plasma samples. The kit includes 12ml of premixed mag beads, 7ml of PBS buffer and an 8 pack of Amicon Ultra centrifugals for downstream buffer exchange or concentration if required.
Application
PureProteome Human Albumin/Immunoglobulin Depletion Kit has been used: to remove specific components from the serum sample for preparing protein stocks using capillary electrophoresis and depletion of all immunoglobulins from plasma.
Features and Benefits
- Matrix comprises of magnetic silica beads coated with polymers and coupled with antibody ligands.
- Beads have specific antibody ligands for recognizing and binding human serum albumin and immunoglobulins.
- Assists in the detection and analysis of proteins of interest.
Components
12 mL of premixed magnetic beads, 7mL of PBS buffer, and an 8-pack of Amicon Ultra-2 mLcentrifugal filters, 3K MWCO
Analysis Note
Depletion: >98% albumin, IgG, IgA, IgM, IgE & IgD depletion. Typical values are >99% depletion.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
IgA and IgM protein primarily drive plasma corona-induced adhesion reduction of PLGA nanoparticles in human blood flow
Daniel J. Sobczynski, Omolola Eniola-Adefeso
Sobczynski DJ and Eniola-Adefeso O
Bioengineering & translational medicine, 2(2), 180-190 (2017)
Daniel J Sobczynski et al.
Bioengineering & translational medicine, 2(2), 180-190 (2017-09-22)
The high abundance of immunoglobulins (Igs) in the plasma protein corona on poly(lactic-co-glycolic) acid (PLGA)-based vascular-targeted carriers (VTCs) has previously been shown to reduce their adhesion to activated endothelial cells (aECs) in human blood flow. However, the relative role of
Semi-permanent cationic coating for protein separations
Crihfield CL, et al.
Journal of Chromatography A, 1607, (2019)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service