Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352202
NACRES:
NA.41
eCl@ss:
32160405
Pricing and availability is not currently available.
biological source
human
Quality Segment
assay
>95% (SDS-PAGE)
form
liquid
manufacturer/tradename
Chemicon®
concentration
4.56 mg/mL protein
General description
LDL constitutes 50% of the total lipoprotein mass in plasma and is the major carrier of cholesterol and cholesteryl esters. LDL levels strongly correlate with coronary heart disease.
Molecular weight = 2,300,000. Da
Composition = 78-80% lipid, 22-20% protein
Density = 1.02 - 1.063 g/mL
Molecular weight = 2,300,000. Da
Composition = 78-80% lipid, 22-20% protein
Density = 1.02 - 1.063 g/mL
MOLECULAR WEIGHT:
2,300,000
DENSITY:
1.02 - 1.063 g/mL
2,300,000
DENSITY:
1.02 - 1.063 g/mL
Product Source: Human plasma tested negative for HBsAg and HIV-I, HIV-II, HBc and Hepatitis C antibodies. All blood products should be treated as potentially infectious.
Physical form
Liquid in 150 mM NaCl, pH 7.4 with 0.01% EDTA. Prepared by sequential flotation ultracentrifugation technique using KBr for density adjustments. Protein is determined by modified Lowry method using BSA as the standard. COMPOSITION: 78-80% lipid, 22-20% protein; CHOLESTROL: 5/44 mg/mL (varies with lot) TRIGLYCERIDE: 1/67 mg/mL (varies with lot)
Preparation Note
Maintain at 2-8ºC in undiluted aliquots for up to 3 months. Do Not Freeze. All blood products should be treated as potentially infectious.
Analysis Note
≥95% of total lipoprotein content as determined by electrophoresis using a SPIFE Cholesterol gel for lipids and SDS-PAGE for proteins.
Other Notes
Replaces: LP2-10MG
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
1 of 1
This Item | |||
|---|---|---|---|
| assay >95% (SDS-PAGE) | assay >95% | assay - | assay ≥95% (of total lipoprotein content, electrophoresis) |
| biological source human | biological source human | biological source human plasma | biological source - |
| form liquid | form liquid | form lyophilized powder | form liquid |
| manufacturer/tradename Chemicon® | manufacturer/tradename Chemicon® | manufacturer/tradename - | manufacturer/tradename Calbiochem® |
| concentration 4.56 mg/mL protein | concentration - | concentration - | concentration - |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



