Skip to Content
Merck
All Photos(3)

Key Documents

IPAKQUAEX

Milli-Q

IPAK Quanta® Polishing Cartridge for Elix® Purified Water

For Milli-Q® IQ/EQ 7000 and IQ 7003 7005 7010 and 7015 ultrapure water systems fed by Elix® pure water

Synonym(s):

IPAK Quanta® cartridge for Elix water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

Milli-Q®

humidity measuring range

10-80% (In original sealed packaging to protect purification elements)

H

39.5 cm (15.5 in.)

input

feed water nature purified water (Elix®)

compatibility

for use with Milli-Q® EQ 7000
for use with Milli-Q® IQ 7000
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015

shipped in

ambient

storage temp.

2-30°C

General description

This IPAK Quanta® polishing cartridge for Elix® water contains innovative IQnano ion-exchange media to remove ions down to trace levels, plus high-grade synthetic activated carbon to target trace organic contaminants.

Combined with an IPAK Meta® water polishing cartridge for Elix® water (IPAKMETEX), it enables the production of ultrapure (Type 1) water for sensitive laboratory applications.

Application

  • For Milli-Q® IQ/EQ 7000 and IQ 7003/05/10/15 ultrapure water systems fed by Elix® water
  • Delivers ultrapure (Type 1) water for sensitive applications

Features and Benefits

  • e-Sure tag enables RFID connection with the system, for seamless consumable status monitoring, data management and full traceability
  • Supplied with a Drain Cap, to remove excess water and reduce the weight of the exhausted cartridge at disposal
  • 78% of the parts suppliers are ISO 14001 environmental management system certified

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK META is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service