Skip to Content
Merck
All Photos(2)

Key Documents

HCD8MAG15K17PMX

Millipore

MILLIPLEX® Human CD8+ T Cell Magnetic Bead Panel Premixed 17 Plex - Immunology Multiplex Assay

Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.

Synonym(s):

Human CD8 T cell cytokine panel, Human cytokine multiplex kit, Luminex® human cytokine immunoassay, Millipore human cytokine panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

standard curve range: 0.4-1,500 pg/mL
(IL-6)

standard curve range: 0.5-2,000 pg/mL
(TNFα)

standard curve range: 1-3,500 pg/mL
(MIP-1α)

standard curve range: 1-5,000 pg/mL
(Granzyme B)

standard curve range: 1-5,000 pg/mL
(IFNγ)

standard curve range: 1-5,000 pg/mL
(IL-5)

standard curve range: 10-50,000 pg/mL
(Perforin)

standard curve range: 10-50,000 pg/mL
(sFasL)

standard curve range: 2-10,000 pg/mL
(IL-4)

standard curve range: 2-10,000 pg/mL
(sCD137)

standard curve range: 2-7,500 pg/mL
(IL-13)

standard curve range: 2-7,500 pg/mL
(IL-2)

standard curve range: 20-100,000 pg/mL
(Granzyme A)

standard curve range: 4-15,000 pg/mL
(GM-CSF)

standard curve range: 400-1,650,000 pg/mL
(sFas)

standard curve range: 5-20,000 pg/mL
(IL-10)

standard curve range: 7-30,000 pg/mL
(MIP-1β)

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

CD8+ T cells (also known as cytotoxic T cells, cytolytic T cells and killer T cells) belong to a sub-group of T cells capable of inducing the death of infected somatic or tumor cells. T cells that bind weakly to Major Histocompatibility Complex (MHC) self-antigens are positively selected into single-positive CD4+ or CD8+ T cells in the thymus. Those CD8+ T cells that survive and mature after activation become cytotoxic cells, expressing T-cell receptors (TCRs) capable of recognizing specific antigenic peptides bound to Class I MHC molecules and the glycoprotein CD8. Affinity between the CD8 protein and the MHC molecule helps to keep the cytotoxic T cell and target closely bound during antigen specific activation. Once activated, CD8+ T cells are generally classified as having a predefined cytotoxic role within the immune system and undergo IL-2 induced clonal expansion, which increases the number of cells specific for the target antigen. The CD8+ T cells then travel throughout the body in search of antigen-positive somatic/tumor cells.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: GM-CSF, sCD137, IFNγ, sFas, sFasL, Granzyme A, Granzyme B, IL-2, IL-4, IL-5, IL-6, IL-10, IL-13, MIP-1α, MIP-1β, TNF-α, Perforin
  • Recommended Sample type: serum, plasma or tissue/cell lysate and culture supernatant
  • Recommended Sample dilution: Neat
  • Assay Run Time: Overnight
  • Research Category: Inflammation & Immunology

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: Refer to kit protocol for sensitivities of individual biomarkers.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sujita Sukumaran et al.
Cancer discovery, 8(8), 972-987 (2018-06-09)
The adoptive transfer of chimeric antigen receptor (CAR)-modified T cells has produced tumor responses even in patients with refractory diseases. However, the paucity of antigens that are tumor selective has resulted, on occasion, in "on-target, off-tumor" toxicities. To address this
Ilana C van Rensburg et al.
Immunity, inflammation and disease, 5(1), 57-67 (2017-03-03)
Studies show that B-cells, in addition to producing antibodies and antigen-presentation, are able to produce cytokines as well. These include regulatory cytokines such as IL-10 by regulatory B-cells. Furthermore, a rare regulatory subset of B-cells have the potential to express
Ariana García-Ojalvo et al.
International wound journal, 16(6), 1294-1303 (2019-08-21)
Diabetic foot ulcer is one of the most frightened diabetic complications leading to amputation disability and early mortality. Diabetic wounds exhibit a complex networking of inflammatory cytokines, local proteases, and reactive oxygen and nitrogen species as a pathogenic polymicrobial biofilm
M Hernandez-Cedeño et al.
Cell stress & chaperones, 26(3), 515-525 (2021-02-26)
Hyperinflammation distinguishes COVID-19 patients who develop a slight disease or none, from those progressing to severe and critical conditions. CIGB-258 is a therapeutic option for the latter group of patients. This drug is an altered peptide ligand (APL) derived from
Lee Kiang et al.
Investigative ophthalmology & visual science, 59(8), 3767-3778 (2018-07-27)
Retinal detachment (RD) separates the retina from the underlying retinal pigment epithelium, resulting in a gradual degeneration of photoreceptor (PR) cells. It is known that RD also results in an inflammatory response, but its contribution to PR degeneration is unknown.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service