HC08
Fluorescent Mounting Media, Aqueous
This Fluorescent Mounting Media, Aqueous is validated for use in Immunofluorescence for the detection of Fluorescent Mounting Media Aqueous.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116133
NACRES:
NA.47
Recommended Products
form
liquid
does not contain
preservative
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
dry ice
storage temp.
−20°C
General description
An aqueous mounting medium designed to preserve and enhance the labeling of immunoreactivity in tissue sections. Sustains the fluorescent signal from labeled samples on slides. Inclusion of 4,6-diamidino-2-phenylindole (DAPI; Cat. No. 268298) allows for the visualization of unlabeled (non-apoptotic) cells. Suitable for use with a wide variety of fluorophores.
Aqueous mounting media designed to preserve and enhance the labeling of tissue sections or cell preparations fixed on slides. Suitable for many fluorophores including fluorescein, Oregon Green, Alexa 488, rhodamine, Texas Red, Cyanine (Cy) 3, and Alexa 546. Inclusion of DAPI in the Fluorescent Mounting Media allows for the visualization of all cells at an excitation wavelength of 330-380 nm.
Application
Immunofluorescence (see comments)
Warning
Toxicity: Standard Handling (A)
Physical form
8 mL in glycerol solution containing 25 mg/ml 1,4-DiAzaBicyclo-[2.2.2]Octane (DABCO) in Tris buffer and 0.1 µg/ml 4,6-DiAmidino-2-PhenylIndole (DAPI).
Reconstitution
Following initial thaw, aliquot and freeze (-20°C).
Other Notes
For best results: following the final aqueous step of the staining procedure remove excess fluid around the specimen, apply 1-2 drops of Fluorescent Mounting Media to the specimen and carefully place a coverglass on top. Seal the border of the coverglass with nail polish. Store mounted specimens at 4°C in the dark.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service