Skip to Content
Merck
All Photos(3)

Key Documents

FCMAB115F

Sigma-Aldrich

Anti-TRA-1-60 Antibody, clone TRA-1-60, FITC conjugate

clone TRA-1-60, from mouse, FITC conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

TRA-1-60, monoclonal

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgM

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PODXL(5420)

General description

Human embryonal carcinoma (EC) cells are the stem cells of teratocarcinomas, and they are key components of germ cell tumors (GCTs). They express several high molecular weight glycoprotein antigens that are down-regulated upon differentiation. One of these antigens, defined by monoclonal antibody TRA-1-60, can be detected in the serum of GCT patients and provides a useful complement to the established serum markers human chorionic gonadotropin and α-fetoprotein, especially in those patients without elevated serum human chorionic gonadotropin or α-fetoprotein.

Specificity

This antibody reacts with TRA-1-60 antigen that is expressed upon the surface of human tetracarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES).
No immunoreactivity is seen with murine EC, EG, or ES cells. Both the TRA-1-60 and TRA-1-81 monoclonal antibodies (MAB4381) recognize antigens that are associated with a pericellular matrix proteoglycan. TRA-1-60 reacts with a sialidase-sensitive epitope whilst TRA-1-81 reacts with an unknown epitope of the same molecule

Immunogen

- Whole Cells corresponding to the of TRA-1-60.

Application

Detect TRA-1-60 using this Anti-TRA-1-60 Antibody, clone TRA-1-60, FITC conjugate validated for use in FC & IC.
Immunocytochemical staining of fixed H9 human ES cells incubated for two hours at 2-8C with 1:100 dilution of anti-TRA-1-60 FITC conjugated (Cat. No. FCMAB115F) monoclonal antibody. Pluripotent human ES cells exhibit strong immunoreactivity to this antibody.
Immunocytochemical staining of live H9 human ES cells incubated for 30 minutes at 37C with 1:100 dilution of anti-TRA-1-60 FITC conjugated (Cat. No. FCMAB115F) monoclonal antibody. Pluripotent human ES cells exhibit strong immunoreactivity to this antibody.
Flow Cytometry:
Antibody dilution for cellular staining:
  • Prepare an antibody working solution by diluting 1:5 the primary antibody with PBS.
  • Dispense the volume per test of working of solution according to the number of cells indicated in the table below.
  • 5 L for Guava Flow Cytometer
  • 10 L for other Flow Cytometry instruments

5 μl Working Solution: 5 x 105 Cells / 95 μl PBS. 100 μl Total Reaction Volume
10 μl Working Solution: 1 x 106 Cells / 90 μl PBS. 100 μl Total Reaction Volume
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Target description

~235/410 kDa

Physical form

Purified mouse monoclonal IgM conjugated to FITC in PBS with less than 0.09% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8ºC protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Pluripotent human embryonic stem (ES) cells

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunbo Yang et al.
Human molecular genetics, 26(16), 3031-3045 (2017-05-19)
Hypoplastic left heart syndrome (HLHS) is among the most severe forms of congenital heart disease. Although the consensus view is that reduced flow through the left heart during development is a key factor in the development of the condition, the
Cathelijn E M Aarts et al.
Stem cell research, 54, 102444-102444 (2021-06-29)
Induced pluripotent stem cells (iPSCs) were generated from blood outgrowth endothelial cells (BOECs) obtained from a healthy donor and from a patient diagnosed with Hermansky Pudlak Syndrome type 2 (HPS2), caused by compound heterozygous AP3B1 mutations (c.177delA and c.1839-1842delTAGA). BOECs
Dario Melguizo-Sanchis et al.
Cell death & disease, 9(2), 128-128 (2018-01-28)
Aplastic Anemia (AA) is a bone marrow failure (BMF) disorder, resulting in bone marrow hypocellularity and peripheral pancytopenia. Severe aplastic anemia (SAA) is a subset of AA defined by a more severe phenotype. Although the immunological nature of SAA pathogenesis
Toshio Miki et al.
Stem cell research & therapy, 7, 8-8 (2016-01-14)
Amnion-derived stem cells have been proposed for cell replacement therapy and tissue regeneration. An easily accessible cell source, the placenta, allows us to potentially establish a bio-bank of cells for immunotype matched clinical applications. Several xeno-free (XF) cryopreservation media are
Irina Neganova et al.
Scientific reports, 7, 41693-41693 (2017-02-06)
Reprogramming of somatic cells to induced pluripotent stem cells (iPSCs) holds enormous promise for regenerative medicine. Reprogramming is a stepwise process with well-defined stages of initiation, maturation and stabilisation which are critically dependent on interactions between key pluripotency transcription factors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service