Skip to Content
Merck
All Photos(3)

Key Documents

EZSTREAM1

Millipore

EZ-Stream® vacuum pump

suitable for bioburden testing, for use with EZ-Fit® Filtration Manifold

Synonym(s):

EZ-Stream® pump, EZ-Stream® vacuum filtration pump

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

Agency

according to ISO 8199

Quality Level

manufacturer/tradename

EZ-Stream®

H

22.2 cm (8.7 in.)

L

16.8 cm (6.6 in.)

W

19.6 cm (7.7 in.)

weight

1.9 kg

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Related Categories

General description

The EZ-Stream® vacuum filtration pump is specifically designed for microbiological analysis. The compact design makes the pump ideal for use on both the work bench and in the laminar air flow hood, where its small footprint saves laboratory space. With the EZ-Stream® pump, the vacuum is provided by a maintenance-free diaphragm and specific check valves that allow liquids to run through the pump. There is no need to use any waste containers normally associated with traditional air pumps or an in-house vacuum. This frees up space in the laboratory working area and eliminates the need for repeated emptying of heavy waste containers. Thanks to its innovative design, the pump has a high-performance flow rate.

Application

EZ-STREAM® vacuum filtration pump is used for microbiology analysis. It is ideal for handling of both liquids and gases, making it suitable for many life science laboratory applications as well.

Features and Benefits

  • Direct transfer ensures no handling of liquid waste
  • High-performance flow rate
  • Noise reduction design
  • Compact
  • Maintenance-free technology
  • Complies with ISO Standard for microbiological analysis

Components

Pump

Multilingual user guide

Power supply

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service