Skip to Content

ES-101

COMPLETE ES CELL MEDIUM W/ 15% FBS AND LIF

Ready to use medium formulated with 15% fetal bovine serum (FBS) and mouse leukemia inhibitory factor (LIF). Each lot is qualified for mouse embryonic stem (ES) cell culture.

Synonym(s):

ES Cell Culture Medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
eCl@ss:
32160801
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Specialty Media

technique(s)

cell culture | mammalian: suitable

input

sample type induced pluripotent stem cell(s)
sample type: mouse embryonic stem cell(s)

General description

Complete ES Cell medium W/ 15% FBS and LIF is suitable for use with mouse embryonic stem (ES) cells. It also works as a complete medium for the growth of primary mouse embryonic fibroblast (PMEF) feeder cells.

Application

Complete ES Cell medium W/ 15% FBS and LIF has been used to culture mouse cranial neural crest cell line (CNCCs). It has also been used to culture mouse embryonic stem cells.

Physical form

Ready to use medium for culture of mouse pluripotent stem (ES and iPS) cells and feeders

Preparation Note

Store at -20°C. After thaw, the medium may be stored at 2-8°C for up to 1 month. Protect from light.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library