CBL517
Anti-CD28 Antibody, clone 15E8
clone 15E8, Chemicon®, from mouse
Synonym(s):
Tp44
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
15E8, monoclonal
species reactivity
human
should not react with
canine
manufacturer/tradename
Chemicon®
technique(s)
flow cytometry: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CD28(940)
Specificity
This antibody reacts with the 44 kDa antigen found on a sub-population of T cells and activated B cells. These CD28+ CD8 cells are now known to be cytotoxic T cell precursors. The antibody does not react with B cells, granulocytes or monocytes. The ligand for CD28 is CD80 (B7) which is found on activated B cells suggesting a role for CD28 in T and B cell interaction
FUSION PARTNER: SP2/0
FUSION PARTNER: SP2/0
Immunogen
Human T lymphocytes
Application
Research Category
Inflammation & Immunology
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
Immunoglobulins & Immunology
This Anti-CD28 Antibody, clone 15E8 is validated for use in FC, FUNC for the detection of CD28.
To study the role of CD28 in T cell proliferation. The antibody will induce the proliferation of resting T lymphocytes.
This antibody will act in cell stimulation studies where it has been suggested that measuring T cell function in HIV infected asymptomatic patients may be an early indicator of AIDS, independent of CD4 + cell counts
Optimal working dilutions must be determined by the end user.
This antibody will act in cell stimulation studies where it has been suggested that measuring T cell function in HIV infected asymptomatic patients may be an early indicator of AIDS, independent of CD4 + cell counts
Optimal working dilutions must be determined by the end user.
Linkage
Replaces: CBL517P
Physical form
Format: Purified
Please see lot specific datasheet for product formulation. We recommend that each laboratory determine an optimum working titre for use in its particular application.
Storage and Stability
For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C for up to one year.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
D Rushworth et al.
Gene therapy, 23(2), 119-128 (2015-08-15)
Antithymidylates (AThy) constitute a class of drugs used in the treatment of cancers such as lung, colon, breast and pancreas. These drugs inhibit DNA synthesis by targeting the enzymes dihydrofolate reductase (DHFR) and/or thymidylate synthase (TYMS). AThys effectively inhibit cancer
Eduardo Huarte et al.
Blood, 112(4), 1259-1268 (2008-06-14)
Robust T-cell responses without autoimmunity are only possible through a fine balance between activating and inhibitory signals. We have identified a novel modulator of T-cell expansion named proliferation-induced lymphocyte-associated receptor (PILAR). Surface PILAR is markedly up-regulated on CD4 and, to
Maria C Lebre et al.
Journal of leukocyte biology, 101(3), 775-783 (2016-10-14)
Bone and cartilage destruction is one of the key manifestations of rheumatoid arthritis (RA). Although the role of T helper (Th)17 cells in these processes is clear, the role of IL-21-producing cells T cells has been neglected. We sought to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service