Skip to Content
Merck

AP112F

Goat Anti-Human IgG Antibody, FITC conjugate

Chemicon®, from goat

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

FITC conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

FITC-conjugated Affinity Purified Goat anti-Human IgG (H+L)

Immunogen

Purified human IgG.

Application

Goat anti-Human IgG Antibody, FITC conjugate is an antibody against Human IgG for use in IF.
Immunohistochemistry: 1:200-1:2000.

Optimal working dilutions must be determined by end user.

DILUTION:

Suggested dilution for most applications: 1:50-1:200

Optimal working dilutions must be determined by the end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Biochem/physiol Actions

Human IgG (H&L)

Physical form

ImmunoAffinity Purified
Lyophilized from 0.02 M phosphate buffered saline, 0.25 M sodium chloride, pH 7.6, with 0.1% sodium azide and 15 mg/mL bovine serum albumin.

Reconstitute with 2 mL of sterile distilled water.

FLUOROPHORE/PROTEIN:

Approximately 14μg/mg

Preparation Note

Maintain lyophilized product at 2-8°C for up to 12 months. After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.

WARNING:

For research use only; not for use as a diagnostic.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library