Skip to Content
Merck
All Photos(1)

Key Documents

AP101

Sigma-Aldrich

Rabbit Anti-Human IgG Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies
This Rabbit anti-Human IgG Antibody is validated for use in ELISA, IP, WB for the detection of Human IgG.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunlong Jia et al.
Cell death & disease, 10(8), 573-573 (2019-08-01)
Emerging evidence suggests that long noncoding RNA (lncRNA) plays pivotal roles in regulating various biological process in human cancers. Titin-antisense RNA1 (TTN-AS1) has been regarded as a tumor promoting lncRNA in numerous cancers. However, the clinical significance and biological function
Yuanyuan Chen et al.
Journal of experimental & clinical cancer research : CR, 42(1), 256-256 (2023-09-30)
Our previous study first showed that ATR-binding long noncoding RNA (lncRNA) is necessary for ATR function and promotes cancer resistance. However, the specific lncRNAs instrumental in ATR activation remain largely unclear, which limits our comprehensive understanding of this critical biological
Yifei Shen et al.
Cell death & disease, 11(4), 278-278 (2020-04-26)
Metabolic switch from oxidative phosphorylation to aerobic glycolysis, which is also called the Warburg effect, is a hallmark of osteosarcoma (OS) and leads to the enhancement of cell chemoresistance, growth, metastasis, and invasion. Emerging evidence indicates that long non-coding RNA
Hui Shen et al.
Cancer science, 114(8), 3128-3143 (2023-05-18)
Many current microRNA (miRNA) expression datasets for renal cell carcinoma (RCC) often show inconsistent analysis results, so a shift to comprehensive analysis of multiple datasets can effectively accelerate molecular screening for precision medicine and translational medicine research. MicroRNA (miR)-188-5p is
Qingke Li et al.
Translational oncology, 46, 101996-101996 (2024-05-26)
Recent studies indicate that circular RNAs (circRNAs) are crucial in the progression of colorectal cancer (CRC). Eukaryotic translation initiation factor 4A3 (EIF4A3) has been identified as a promoter of circRNA production. The biological roles and mechanisms of EIF4A3-derived circRNA (circEIF4A3)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service