Skip to Content
Merck

AM63

Anti-A20 Mouse mAb (59A426)

liquid, clone 59A426, Calbiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
59A426, monoclonal
Species reactivity:
human
Application:
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

59A426, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

dilution

(Immunoblotting (1:500-1:2000)
Immunoprecipitation (2 µg/mg))

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNFAIP3(7128)

General description

Anti-A20 mouse monoclonal antibody, clone 59A426, recognizes ~70 kDa A20 protein in Daudi cells. It is validated for use in immunoblotting and immunoprecipitation.
Protein G purified mouse monoclonal antibody. Recognizes the ~70 kDa A20 protein.
Recognizes the ~70 kDa A20 protein in Daudi cells.

Immunogen

Human
full-length recombinant human A20 protein

Application


Immunoblotting (1:500-1:2000)
Immunoprecipitation (2 g/mg)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In PBS, 0.2% gelatin.

Preparation Note

Following initial thaw, aliquot freeze (-20°C).

Analysis Note

Positive Control
Daudi cells

Other Notes

Antibody should be titrated for optimal results in individual systems.
Heyninck, K., et al. J. Cell Biol145, 1471.
May, M.J. et al. 1997. Seminars in Cancer Biology.2, 63.
Tewari, M., et al. 1995. J. Immunol.154, 1699.
Opipari, A.W., et al. 1990. J. Biol. Chem.265, 14705.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library