Skip to Content
Merck

Skip To

AG970

β Amyloid 1-42, aβ, ultra pure, NaOH, recombinant human

Synonym(s):

β Amyloid 1-42, Recombinant Amyloid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.41
eCl@ss:
32160405
Form:
powder
Assay:
>97.0%
Biological source:
human
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

assay

>97.0%

form

powder

manufacturer/tradename

Chemicon®

NCBI accession no.

UniProt accession no.

Gene Information

human ... APP(351)

General description

Beta Amyloid (1-42), ultra pure, NaOH

MOL. WT.: 4514.3 Da by Mass Spec Analysis (theoretical 4514.10)

SEQUENCE: H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH

PEPTIDE CONTENT: 500 μg per vial.

COUNTER ION: NaOH

Application

Different researchers claim enhanced activity due to different salts.



The TFA salt is what has been the industry standard.



The HFIP makes the beta-amyloid homogenously monomeric.



The NaOH apparently make it monomeric too and is also water soluble, which the TFA salt and HFIP are not.



The HCl salt is supposed to be more biological, as TFA is not found in the body.



By offering the different salts, it lets the researchers duplicate the different assays that have been developed by other researchers (see below).



TFA: Shu-chuan Jao, S-C, et. al. 1997, Amyloid; Int J Exp. Clinical Invest. 4, 231

HFIP: Stine, W.B. et. al. 2003, J. Biol. Chem, 278, 11612

NaOH: Fezoui, Y., et. al. 2000, Amyloid, 7, 166

HCL: Kanek, I and Tutumi, S., 1997, J. Neurochem, 68, 438
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Physical form

White lyophilized powder. Resuspend in any one of the following solvents: 0.1% TFA, 5 mM HCl, 2 mM NaOH, 0.2% NH4OH at a concentration of 1 mg/mL. Sonicate for 30 seconds to 1 minute after it has gone into solution. To bring into your solution: After resuspension, add 5x or 10x buffer stock (PBS, TBS) and water to bring to 1x buffer.

Preparation Note

Maintain lyophilized material at -20ºC for up to 12 months after date of receipt. After reconstitution maintain at -20ºC to -70ºC for up to 2 weeks in undiluted aliquots. Avoid freeze/thaw cycles to avoid aggregation.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AG912AG972AG964
biological source

human

biological source

human

biological source

human

biological source

human

assay

>97.0%

assay

>97%

assay

>97.0%

assay

98%

form

powder

form

powder

form

powder

form

powder

UniProt accession no.

P05067

UniProt accession no.

P05067

UniProt accession no.

P05067

UniProt accession no.

P05067

Gene Information

human ... APP(351)

Gene Information

human ... APP(351)

Gene Information

human ... APP(351)

Gene Information

human ... APP(351)

NCBI accession no.

NM_000484.2, NM_201414.1, NM_201413.1

NCBI accession no.

NM_000484.2, NM_201413.1, NM_201414.1

NCBI accession no.

NM_000484.2, NM_201413.1, NM_201414.1

NCBI accession no.

NM_000484.2, NM_201413.1, NM_201414.1


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters