Skip to Content
Merck

AG910

β-Amyloid Peptide

98%, powder, 1-40, human recombinant

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

β Amyloid 1-40, aβ, ultra pure, TFA, recombinant human,

biological source

human

Quality Level

assay

98%

form

powder

manufacturer/tradename

Chemicon®

NCBI accession no.

UniProt accession no.

Gene Information

human ... APP(351)

General description

Beta Amyloid (1-40), ultra pure, TFA

MOL. WT.: 4329.6 Da by Mass Spec Analysis (theoretical 4329.86)

SEQUENCE: Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val

PEPTIDE CONTENT: 1 mg per vial; salt content is <10%

COUNTER ION: TFA

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
AG910-1MG04053252621147