Skip to Content
Merck
All Photos(1)

Key Documents

ABS524

Sigma-Aldrich

Anti-Lipin-2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Phosphatidate phosphatase LPIN2, Lipin-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
IP
WB
species reactivity:
mouse
technique(s):
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Lpin2(64898)

General description

Lpin2, also known as Phosphatidate phosphatase LPIN2, and encoded by the gene Lpin2, plays important roles in controlling the metabolism of fatty acids. Lpin2 acts as a magnesium-dependent phosphatidate phosphatase enzyme which catalyzes the conversion of phosphatidic acid to diacylglycerol during triglyceride, phosphatidylcholine and phosphatidylethanolamine biosynthesis in the reticulum endoplasmic membrane. Lpin2 also acts as a nuclear transcriptional coactivator for PGC-1α protein. Lpin2 is localized to the cytoplasm and endoplasmic reticulum membrane but is also found in the nucleus. Lpin2 is expressed in many cells and tissues including liver, lung, kidney, placenta, prostate, and intestine. Expression is also seen in RBCs and at sites of lymphopoiesis in bone and spleen. Homozygous mutations in LPIN2 result in Majeed syndrome. Majeed syndrome is an autosomal recessive, autoinflammatory disorder characterized by chronic recurrent multifocal osteomyelitis and congenital dyserythropoietic anemia.

Immunogen

Linear peptide corresponding to mouse Lipin-2.

Application

Anti-Lipin-2 Antibody is an antibody against Lipin-2 for use in western blotting, IHC & IP.
Research Category
Signaling
Research Sub Category
Lipid Metabolism & Weight Regulation
Western Blotting Analysis: A representative lot detected Lipin-2 in mouse WAT cell lysate (Gropler, M. C., et al. (2008). JBC. 284(11):6763-6772).
Immunocytochemistry Analysis: A representative lot detected Lipin-2 in mouse cerebellum tissue (Mayurranjan, S., et al. (2013). PNAS. 110(2):642-647).

Quality

Evaluated by Western Blotting in mouse hepatocytes cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Lipin-2 in 10 µg of mouse hepatocytes cell lysate.

Target description

~ 110 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide with 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingquan He et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 31(7), 2893-2904 (2017-03-30)
Cancer cells reprogram their metabolism to increase the synthesis of macromolecules for rapid proliferation. Compared to fatty acids, much less is known about the synthesis of phospholipids, which is essential for membrane biogenesis in cancer cells. We found that LPIN1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service