Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IP, WB
Citations:
-
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
mol wt
calculated mol wt 112.46 kDa, observed mol wt ~42 kDa
purified by
affinity chromatography
species reactivity
mouse
packaging
antibody small pack of 100 μg
technique(s)
immunofluorescence: suitable, immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG
epitope sequence
C-terminal half
Protein ID accession no.
UniProt accession no.
shipped in
2-8°C
target post-translational modification
phosphorylation (pSer746)
General description
Serine/threonine-protein kinase ULK1 (UniProt: O70405; also known as EC:2.7.11.1, Serine/threonine-protein kinase Unc51.1, Unc-51-like kinase 1) is encoded by the Ulk1 gene (Gene ID: 22241) in murine species. ULK-1 is a cytosolic serine/threonine protein kinase that is localized in cytosol and preautophagosomal structures. Under condition of starvation, it is localized to punctate structures primarily representing the isolation membrane that sequesters a portion of the cytoplasm resulting in the formation of an autophagosome. Its protein kinase domain is localized to amino acids 16-278. It is involved in autophagy in response to starvation and acts upstream of PI3-kinase (PIK3C3) to regulate the formation of autophagophores, the precursors of autophagosomes. It acts both as a downstream effector and a negative regulator of mTORC1 via interaction with Raptor. ULK1 contains multiple phosphorylation sites, and their phosphorylation status is reported to regulate canonical autophagy. Under unstimulated conditions, it is phosphorylated at serine 637 and 757 by mTOR complex that leads to its inactivation. ULK1 is activated via phosphorylation by AMPK and also acts as a negative regulator of AMPK through phosphorylation of the AMPK subunits PRKAA1, PRKAB2 and PRKAG1. Despite its similarity with ULK2 in its enzymatic domain, there are major differences in their autophagy-related interactors and their post-translational and transcriptional regulators. ULK1 undergoes phosphorylation at serine 746 by receptor interacting protein kinase 3 (RIPK3) during genotoxic stress-induced alternative autophagy and this phosphorylated form is shown to localize exclusively on the Golgi and is essential for alternative autophagy, but not canonical autophagy. (Ref.: Torii, S., et al. (2020). Nat. Commun. 11: 1754).
Immunogen
KLH-conjugated linear peptide corresponding to 16 amino acids surrounding phosphoserine 746 of mouse ULK-1.
Application
Quality Control Testing
Evaluated by Western blotting with constructs of ULK-1 phosphorylated on Serine 746.
Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected MBP-tagged ULK1 construct phosphorylated on serine 746, but not in non-phosphorylated construct.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected ULK1 phosphorylated on serine 746.
Immunoprecipitation Analysis: A representative lot immunoprecipitated phospho ULK1(Ser746) in Immunoprecipitation applications (Satoru Torii, et al. (2020). Nat. Commun. 11(1):1754).
Immunofluorescence Analysis: A representative lot detected phospho ULK1(Ser746) in Immunofluorescence applications (Satoru Torii, et al. (2020). Nat. Commun. 11(1):1754).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Evaluated by Western blotting with constructs of ULK-1 phosphorylated on Serine 746.
Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected MBP-tagged ULK1 construct phosphorylated on serine 746, but not in non-phosphorylated construct.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected ULK1 phosphorylated on serine 746.
Immunoprecipitation Analysis: A representative lot immunoprecipitated phospho ULK1(Ser746) in Immunoprecipitation applications (Satoru Torii, et al. (2020). Nat. Commun. 11(1):1754).
Immunofluorescence Analysis: A representative lot detected phospho ULK1(Ser746) in Immunofluorescence applications (Satoru Torii, et al. (2020). Nat. Commun. 11(1):1754).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-phospho ULK1(Ser746), Cat. No. ABS2250, is a rabbit polyclonal antibody that detects ULK1 phosphorylated on serine 746 and is tested for use in Immunofluorescence, Immunoprecipitation, and Western Blotting.
Biochem/physiol Actions
This rabbit polyclonal antibody detects ULK-1 phosphorylated on serine 746.
Physical form
Purified rabbit polyclonal antibody in buffer containing PBS with 0.09% sodium azide
Preparation Note
Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.