Skip to Content
Merck
All Photos(1)

Key Documents

AB9890

Sigma-Aldrich

Anti-AGE (Advanced Glycation End-products) Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

goat

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

AGE (Advanced Glycation End-products)

Immunogen

Glycoaldehyde-modified protein.

Application

Anti-AGE (Advanced Glycation End-products) Antibody detects level of AGE (Advanced Glycated End-products) & has been published & validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Oxidative Stress
Western blot: ≥1:2,000 on AGE-modified proteins.

ELISA: ≥ 1:4,000 against Glycoaldehyde-modified BSA.

Optimal working dilutions must be determined by end user.

Physical form

Goat antiserum. Liquid with 0.1% SODIUM AZIDE
Serum

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shweta Kishor Sonawane et al.
Oncotarget, 12(11), 1083-1099 (2021-06-05)
Alzheimer's disease is a type of dementia denoted by progressive neuronal death due to the accumulation of proteinaceous aggregates of Tau. Post-translational modifications like hyperphosphorylation, truncation, glycation, etc. play a pivotal role in Tau pathogenesis. Glycation of Tau aids in
Vassilis L Tzounakas et al.
Frontiers in medicine, 5, 16-16 (2018-02-21)
Transfusion of fresh frozen plasma (FFP) helps in maintaining the coagulation parameters in patients with acquired multiple coagulation factor deficiencies and severe bleeding. However, along with coagulation factors and procoagulant extracellular vesicles (EVs), numerous bioactive and probably donor-related factors (metabolites
Ali S Alqahtani et al.
Saudi journal of biological sciences, 28(1), 813-824 (2021-01-12)
Current pre-clinical evidences of Centella focus on its pharmacological effects on normal wound healing but there are limited studies on the bioactivity of Centella in cellular dysfunction associated with diabetic wounds. Hence we planned to examine the potential of Centella
Amit Garg et al.
ACS pharmacology & translational science, 6(2), 253-269 (2023-02-18)
Advanced glycation end-products (AGEs) form when glucose reacts non-enzymatically with proteins, leading to abnormal protein function, oxidative stress, and inflammation. AGEs are associated with aging and age-related diseases; their formation is aggravated during diabetes. Therefore, drugs preventing AGE formation can
Hugo Vicente Miranda et al.
Brain : a journal of neurology, 140(5), 1399-1419 (2017-04-12)
α-Synuclein misfolding and aggregation is a hallmark in Parkinson's disease and in several other neurodegenerative diseases known as synucleinopathies. The toxic properties of α-synuclein are conserved from yeast to man, but the precise underpinnings of the cellular pathologies associated are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service