Skip to Content
Merck
All Photos(1)

Key Documents

AB9179

Sigma-Aldrich

Anti-Glucagon-Like Peptide 2 Receptor Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
species reactivity:
human
technique(s):
immunohistochemistry: suitable (paraffin)
citations:
2

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

availability

not available in Japan

technique(s)

immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GLP2R(9340)

Specificity

Recognizes Glucagon-Like Peptide 2 Receptor.

Immunogen

Synthetic peptide from the N-terminal extracellular domain of human Glucagon-Like Peptide 2 Receptor.

Application

Anti-Glucagon-Like Peptide 2 Receptor Antibody is an antibody against Glucagon-Like Peptide 2 Receptor for use in IH(P).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K K Qi et al.
Animal : an international journal of animal bioscience, 9(9), 1481-1489 (2015-05-13)
This study was conducted to determine the effects on intestinal function, anti-inflammatory role and possible mechanism of polyethylene glycosylated (PEGylated) porcine glucagon-like peptide-2 (pGLP-2), a long-acting form of pGLP-2, in weaning piglets challenged with Escherichia coli lipopolysaccharide (LPS). We divided
K K Qi et al.
Journal of animal physiology and animal nutrition, 101(6), 1242-1248 (2017-02-06)
Because of rare glucagon-like peptide-2 (GLP-2) receptor (+) cells within the gut mucosa, the molecular mechanisms transducing the diverse actions of GLP-2 remain largely obscure. This research identified the naturally occurring intestinal cell lines that endogenously express GLP-2R and determined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service