Skip to Content
Merck
All Photos(1)

Key Documents

AB911

Sigma-Aldrich

Anti-Oxytocin Antibody

serum, Chemicon®

Synonym(s):

Anti-OT, Anti-OT-NPI, Anti-OXT-NPI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat, rabbit, sheep, hamster

manufacturer/tradename

Chemicon®

technique(s)

dot blot: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
radioimmunoassay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... OXTR(5021)

Specificity

Recognizes Oxytocin. Less than 1% cross-reactivity with arginine vasopressin.

Immunogen

Synthetic oxytocin (Sigma) conjugated to thyroglobulin.

Application

Anti-Oxytocin Antibody is an antibody against Oxytocin for use in DB, IC, IH(P), RIA.
Immunohistochemistry: 1:5,000-1:10,000 (18-24 hour incubation) on formalin, Bouin′s or Zamboni′s fixed tissue. Will react on paraffin embedded tissue.

Immunocytochemistry: 1:10,000 on fetal rat neurons.

Dot Blot: 1:1000

RIA

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Hormones & Receptors

CNS Control of Metabolism

Physical form

Neat rabbit antisera. Liquid. Contains no preservative.
Unpurified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Pituitary Gland

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole Denoix et al.
Frontiers in neurology, 11, 649-649 (2020-08-06)
The hydrogen sulfide (H2S) and the oxytocin/oxytocin receptor (OT/OTR) systems interact in trauma and are implicated in vascular protection and regulation of fluid homeostasis. Acute brain injury is associated with pressure-induced edema formation, blood brain barrier disruption, and neuro-inflammation. The
Yu-Ting Lin et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(5), 1218-1231 (2017-12-28)
Oxytocin (OXT) receptors (OXTRs) are prominently expressed in hippocampal CA2 and CA3 pyramidal neurons, but little is known about its physiological function. As the functional necessity of hippocampal CA2 for social memory processing, we tested whether CA2 OXTRs may contribute
Christine N Yohn et al.
Frontiers in behavioral neuroscience, 12, 181-181 (2018-09-14)
Natural variations in parenting are associated with differences in expression of several hormones and neuropeptides which may mediate lasting effects on offspring development, like regulation of stress reactivity and social behavior. Using the bi-parental California mouse, we have demonstrated that
Karin Warfvinge et al.
The journal of headache and pain, 21(1), 10-10 (2020-02-08)
Recent work, both clinical and experimental, suggests that the hypothalamic hormone oxytocin (OT) and its receptor (OTR) may be involved in migraine pathophysiology. In order to better understand possible central actions of OT in migraine/headache pathogenesis, we mapped the distribution
Mehwish Anwer et al.
Journal of neurotrauma, 37(7), 924-938 (2019-10-28)
Traumatic brain injury (TBI) causes damage to the hypothalamo-hypophyseal axis, leading to endocrine dysregulation in up to 40% of TBI patients. Hence, there is an urgent need to identify non-invasive biomarkers for TBI-associated hypothalamo-hypophyseal pathology. Sushi repeat-containing protein X-linked 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service