Skip to Content
Merck
All Photos(3)

Documents

AB6046

Sigma-Aldrich

Anti-Cytoplasmic FMR1-interacting protein 1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

cytoplasmic FMR1 interacting protein 1, Specifically Rac1-associated protein 1, cytoplasmic FMRP interacting protein 1, cytoplasmic FMR1-interacting protein 1, selective hybridizing clone

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rat (based on 100% sequence homology), primate (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SRA1(10011)

General description

Cytoplasmic FMR1-interacting protein 1 (CYFIP1) is a member of the CYFIP family. It is a component of the CYFIP1-EIF4E-FMR1 complex, a regulatory complex for translational repression that binds to the mRNA cap. Within the complex, CYFIP1 serves as an adapter between the other two components and upregulates the translation repression activity of FMR1. It regulates epithelial morphogenesis, binds F-actin, and is a component in the development and formation of axon outgrowth, lamellipodia, protrusions, and membrane ruffles. Through its interaction with RAC1, CYFIP1 regulates the activity of WASP family members. CYFIP1 is associated with Prader-Willi/Angelman syndromes and may also have a role in autism.

Immunogen

Epitope: Unknown
KLH-conjugated linear peptide corresponding to an internal sequence (a.a.519-532) of human Cytoplasmic FMR1-interacting protein 1.

Application

Detect Cytoplasmic FMR1-interacting protein 1 using this Anti-Cytoplasmic FMR1-interacting protein 1 Antibody validated for use in WB & IC.
Research Category
Cell Structure

Signaling
Research Sub Category
Cytoskeletal Signaling
Western Blot (Snap i.d.) Analysis: 5 µg/mL from a previous lot detected Cytoplasmic FMR1-interacting protein 1 in 10 µg of HeLa cell lysate.
Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected Cytoplasmic FMR1-interacting protein 1 in NIH/3T3, HeLa, and A431 cells.

Quality

Evaluated by Western Blot in HeLa cell lysate.
Western Blot Analysis: 1 µg/mL of the antibody detected Cytoplasmic FMR1-interacting protein 1 on 10 µg of HeLa cell lysate.

Target description

~ 154 kDa observed. There are three noted isoforms with calculated MWs of 51 kDa, 94 kDa, and 145 kDa Additional isoforms may be observed in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gilles Gouspillou et al.
Scientific reports, 5, 8717-8717 (2015-03-04)
Anticancer treatments for childhood acute lymphoblastic leukaemia (ALL) are highly effective but are now implicated in causing impaired muscle function in long-term survivors. However, no comprehensive assessment of skeletal muscle mitochondrial functions in long-term survivors has been performed and the
Agustin Gonzalez-Vicente et al.
Nutrients, 9(8) (2017-08-17)
Fructose-enriched diets cause salt-sensitive hypertension. Proximal tubules (PTs) reabsorb 70% of the water and salt filtered through the glomerulus. Angiotensin II (Ang II) regulates this process. Normally, dietary salt reduces Ang II allowing the kidney to excrete more salt, thereby
Surya Nagaraja et al.
Cancer cell, 31(5), 635-652 (2017-04-25)
Diffuse intrinsic pontine glioma (DIPG) is a fatal pediatric cancer with limited therapeutic options. The majority of cases of DIPG exhibit a mutation in histone-3 (H3K27M) that results in oncogenic transcriptional aberrancies. We show here that DIPG is vulnerable to
Heba Elsayed et al.
Frontiers in bioengineering and biotechnology, 8, 784-784 (2020-08-06)
Following peripheral nerve injury, a sequence of events termed Wallerian degeneration (WD) takes place at the distal stump in order to allow the regenerating axons to grow back toward the target organs. Schwann cells (SCs) play a lead role in
Ruiying Ma et al.
Animal cells and systems, 27(1), 93-101 (2023-04-01)
Cytoplasmic FMR1-interacting protein 2 (CYFIP2) is an evolutionarily conserved multifunctional protein that regulates the neuronal actin cytoskeleton, mRNA translation and transport, and mitochondrial morphology and function. Supporting its critical roles in proper neuronal development and function, human genetic studies have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service