Skip to Content
Merck

AB6000

Anti-TIMP-3 Antibody, CT

from rabbit, purified by affinity chromatography

Synonym(s):

MIG-5, TIMP metallopeptidase inhibitor 3, Tissue inhibitor of metalloproteinases 3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
13
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

horse, human, rat

species reactivity (predicted by homology)

pig (based on 100% sequence homology), primate (based on 100% sequence homology), equine (based on 100% sequence homology), canine (based on 100% sequence homology), monkey (based on 100% sequence homology), mouse (95% homology), bovine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TIMP3(7078)

General description

Bands may be observed for reduced protein bands at ~ 24 kDa (unglycosylated) and ~ 30 kDa (glycosylated). Additional TIMP-3 bands MAY be visible at 50 kDa (dimer), 12 kDa, and 15 kDa (breakdown products) and sample dependent.
Tissue inhibitor of metalloproteinases 3 (TIMP-3) is a member of the TIMP family. The TIMPs are so named because they are slow, tight binding endogenous inhibitors of the Matrix Metalloproteinases (MMPs). The four TIMPs have different inhibition constants for the different MMPs studied. TIMPs have been shown to have activity against the ADAMs family of proteinases. TIMP-3 is an efficient "sheddase" inhibitor, inhibiting ADAM-17 (TACE) at the low nanomolar levels seen with MMP inhibition. The other ADAMs proteinases have not yet been assayed for TIMP inhibition, but TIMP-2 seems to be much less active on ADAM-17 than isTIMP-3. TIMP-3 is thought to be constitutively produced by many cell types and is inducible in others. The TIMP-3 localization differs from the other 3 TIMPs, and is thought to be primarily deposited into the extracellular matrix.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to human TIMP-3 at the C-terminus.

Application

Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected TIMP-3 in C6 cells.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
This Anti-TIMP-3 Antibody, C-terminus is validated for use in WB, IC for the detection of TIMP-3.

Biochem/physiol Actions

The antibody recognizes TIMP-3 at the C-terminus. Does not appear to cross react with TIMP-1 or TIMP-2.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HL-60 cell lysate in PMA-conditioned media.
Evaluated by Western Blot using an HL-60 cell lysate supplemented with PMA-conditioned media.

Western Blot Analysis: 0.5 µg/ml of this antibody detected TIMP-3 on 10 µg of HL-60 in PMA-conditioned media.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: AB802

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
AB600004053252279706