Skip to Content
Merck
All Photos(2)

Key Documents

AB3212-I

Sigma-Aldrich

Anti-Na+/HCO3- Cotransporter Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Electrogenic sodium bicarbonate cotransporter 1, Sodium bicarbonate cotransporter, Na(+)/HCO3(-) cotransporter, Solute carrier family 4 member 4, kNBC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
WB
species reactivity:
rat, human
technique(s):
immunohistochemistry: suitable
western blot: suitable
citations:
4

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

species reactivity (predicted by homology)

rabbit (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC4A4(8671)

General description

Electrogenic sodium bicarbonate cotransporter 1 (UniProt Q9Y6R1; also known as Sodium bicarbonate cotransporter, Na(+)/HCO3(-) co-transporter, Solute carrier family 4 member 4, kNBC1) is encoded by the SLC4A4 (also known as NBC, NBC1, NBCE1) gene (Gene ID 8671) in human. Bicarbonate (HCO3-) transporters are the principal regulators of pH in cells and play a vital role in acid-base movement in various tissues. There exist four types of HCO3- transporters, namely Cl-/-HCO3- exchangers, Na+/HCO3- co-transporters (NBCs), a K+/HCO3- co-transporter, and a Na+-driven Cl-/-HCO3- exchanger. SLC4A4 is a multi-pass membrane NBC with 10 transmembrane segments, 5 extracellular domains, and 6 intracellular domains, having both its N- and C-terminal (a.a. 1-468 and a.a. 987-1079, respectively) exposed intracellularly. NBCs mediate the coupled movement of sodium and bicarbonate ions, in a 3 to 1 stoichiometric ratio, across the plasma membrane. Multiple isoforms are reported as a result of alternative splicing, including NBCe1-A (kNBC1, kNBC-1; UniProt Q9JI66-2/Rat and Q9Y6R1-2/Human) that is mainly expressed in the kidney, NBCe1-B (bNBC1, pNBC-1; UniProt Q9JI66-1/Rat and Q9Y6R1-1/Human) expressed in the pancreas and other tissues, and NBCe1-C (b2NBC; UniProt Q9JI66-3/Rat and Q9Y6R1-5/Human) expressed in the brain.

Specificity

Expected to react with all rat and human spliced isoforms reported by UniProt (Q9JI66 and Q9Y6R1, respectively).

Immunogen

Epitope: Amino acids 338-391
GST-tagged recombinant protein corresponding to amino acids 338-391 of human Na+/HCO3- cotransporter.

Application

Detect Na+/HCO3- Cotransporter using this Anti-Na+/HCO3- Cotransporter Antibody validated for use in Western Blotting, Immunohistochemistry.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Na+/HCO3- cotransporter in human cerebral cortex, kidney and pancreas tissues.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Quality

Evaluated by Western Blotting in human kidney tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Na+/HCO3- cotransporter in 10 µg of human kidney tissue lysate.

Target description

~130 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Štefan Baňas et al.
International journal of molecular sciences, 25(7) (2024-04-13)
Kaempferol (KAE) is a natural flavonoid with powerful reactive oxygen species (ROS) scavenging properties and beneficial effects on ex vivo sperm functionality. In this paper, we studied the ability of KAE to prevent or ameliorate structural, functional or oxidative damage
Filip Benko et al.
International journal of molecular sciences, 23(23) (2022-12-12)
Since the molecular similarities and differences among physiological capacitation and cryocapacitation have not been studied in detail, this study was designed to assess the gene and protein expression levels of the Cation channel of sperm (CatSper) 1 and 2, sodium
Cary R Boyd-Shiwarski et al.
American journal of physiology. Renal physiology, 318(6), F1341-F1356 (2020-04-14)
We characterized mouse blood pressure and ion transport in the setting of commonly used rodent diets that drive K+ intake to the extremes of deficiency and excess. Male 129S2/Sv mice were fed either K+-deficient, control, high-K+ basic, or high-KCl diets
Štefan Baňas et al.
International journal of molecular sciences, 24(3) (2023-02-12)
Epicatechin (EPC) is a flavonoid belonging to the family of catechins; it has been described as a powerful scavenger of a wide spectrum of reactive oxygen species (ROS) and a modulator of ex vivo sperm vitality. In this study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service