Skip to Content
Merck
All Photos(1)

Key Documents

AB3200

Sigma-Aldrich

Anti-LIM-1 Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-Anti-LIM-1, Anti-Anti-LIM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
IF
IHC
IP
WB
species reactivity:
human, frog, fish, mouse
technique(s):
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable
citations:
4

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, frog, fish, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LHX1(3975)

Specificity

Recognizes LIM-1. This antibody does not appear to cross react with LIM-5 (the closest homologue of LIM-1 in frog) on tissue sections but does cross react with LIM-5 in Western blot and immunoprecipitation.

Immunogen

C-terminal portion of frog LIM-1 protein.

Application

Anti-LIM-1 Antibody detects level of LIM-1 & has been published & validated for use in IF, IP, WB, IC, IH(P).
Immunohistochemistry: 1:200-1:1,000 (Fish and whole mounts 1:500) Recommended fixative MEMFA.

Our photo was produced from immersion fixation of chicken embryos in cold 4% PFA for 15-30 minutes, then sectioning was performed on a cryostat.

Works on paraffin embedded sections when sections are either lightly PFA fixed or fixed with acetone, ethanol or fixative suggested below.

SUGGESTED IMMUNOHISTOCHEMISTRY PROTOCOL FOR AB3200

The best fixative is MEMFA. 10X stock solution for MEMFA: 1 M MOPS, 20 MM EGTA. 10 mM MgSO4, 38% Formaldehyde. Fixation 1 hour, 2 x 15 min methanol. Following this protocol embryos may be stored in methanol at -20°C indefinitely or immediately embedded in paraplast. Best results on paraffin sections 6-10 micron thick.2) Staining following deparaffinization in xylene and a row of alcohol wash two times in water. Block in 2% Boehringer Mannheim reagent in 0.1 M maleic acid, pH 7.5, 150 mM NaCl for one hour at room temperature.3) Dilute the AB3200 in same blocking reagent and incubate overnight at 4°C or for at least 5 hours. Wash three times in PBS, 10 min each.4) Incubate with alkaline phosphatase conjugated secondary antibody (for example Chemicon Catalog Number AP132A. Develop with BCIP/TNBT (Chemicon Catalog Number ES007-100ML).5) For sections always use Digene silanated slides or Superfrost plus from Fisher as some times you may need to boil sections in 6 M urea for 5-6 min. in microwave at 80% power following deparaffinization to increase signal. That is especially useful if tissues were fixed in PFA.6) Sometimes it is necessary to predeplete antibody on hyperfixed embryos to lower background (especially for staining species other than frog and for whole-mounts). Procedure: hyperfix frog or fish embryos in MEMFA for 36-48 hours at RT 30-50 embryos. Wash 2 X in methanol (see above). Apply antibody in final dilution in blocking reagent for one hour on rocking table. Collect super and apply to your embryos or sections. For whole mounts use the following procedure: after fixation, methanol and PBS; block in PBST + serum (PBS + 2 mg/mL BSA + 0.1% Triton X100 + 10% animal serum) one hour room temperature. Add first antibody in PBST + serum and incubate over night at 4°C. Wash in PBST (no serum) 4 times for 2 hours. Add secondary diluted in PBST + serum over night at 4°C. Wash in PBST four times for 2 hours. Develop staining.

Do not use tissues fixed overnight in PFA the antibody will not work.

Immunocytochemistry: 1:500 on P19 cell line, lightly fixed (2% PFA) 5-15 minutes, permeabilized with 0.1% triton X-100 or methanol ( 5′ air dry).

Western blot: 1:3,000-1:6,000

Immunoprecipitation: 1:200

Immunofluorescence 1:100

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Neuronal & Glial Markers

Physical form

Format: Purified
Purified immunoglobulin

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiri Pergner et al.
Frontiers in cell and developmental biology, 8, 705-705 (2020-08-28)
The evolution of the vertebrate eye remains so far unresolved. Amphioxus frontal eye pigment cells and photoreceptors were proposed to be homologous to vertebrate photoreceptors and retinal pigmented epithelium, based on ultrastructural morphology and gene expression analysis in B. floridae.
Onecut1 is essential for horizontal cell genesis and retinal integrity.
Wu, F; Li, R; Umino, Y; Kaczynski, TJ; Sapkota, D; Li, S; Xiang, M; Fliesler, SJ; Sherry et al.
The Journal of Neuroscience null
Keng Ioi Vong et al.
Molecular brain, 8, 25-25 (2015-04-19)
The high mobility group (HMG) family transcription factor Sox9 is critical for induction and maintenance of neural stem cell pool in the central nervous system (CNS). In the spinal cord and retina, Sox9 is also the master regulator that defines
Ka Kui Tong et al.
Molecular and cellular biology, 33(10), 1925-1937 (2013-03-06)
Bone morphogenetic protein (BMP) signaling is critical for cerebellum development. However, the details of receptor regulated-Smad (R-Smad) and common partner Smad (Co-Smad, or Smad4) involvement are unclear. Here, we report that cerebellum-specific double conditional inactivation of Smad1 and Smad5 (Smad1/5)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service