Skip to Content
Merck

AB2204

Anti-BMAL1 Antibody

serum, from guinea pig

Synonym(s):

ARNT-like protein 1, brain and muscle, Basic-helix-loop-helix-PAS protein MOP3, Brain and muscle ARNT-like 1, member of PAS protein 3, aryl hydrocarbon receptor nuclear translocator-like, bHLH-PAS protein JAP3, basic-helix-loop-helix-PAS orphan MOP3 2, m

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
8
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

guinea pig

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ARNTL(406)
mouse ... Arntl(11865)
rat ... Arntl(29657)

General description

Brain and muscle Arnt-like protein-1 (BMAL1; also known as MOP3 or Arnt3) is a transcription factor known to regulate circadian rhythm. BMAL1 is the only component of the mammalian circadian clock whose sole deletion in a mouse model generates arrhythmicity. In addition to defects in the clock, these BMAL1 null-mice also have reproductive problems are small in stature, age quickly and have progressive arthropathy that results in having less overall locomotor activity than wild type mice. Recent phenotyping data suggests that BMAL1 and its partner Clock also play a role in regulation of glucose homeostasis and metabolism. Finally, BMAL1, NPAS2, and PER2 have been associated with seasonal affective disorder in humans. BMAL1 transcription is circadian and reciprocally regulated by NR1D1 (Rev-erb-alpha) and RORA, which establishes a second interlocking loop in the mammalian circadian clock.
~69 kDa

Immunogen

Epitope: C-Terminus
Recombinant Protein

Application

Anti-BMAL1 Antibody is an antibody against BMAL1 for use in WB.
Research Category
Neuroscience
Research Sub Category
Circadian Rhythm & Sleep

Biochem/physiol Actions

Cat. # AB2204 will recognize the C-terminus of BMAL1.
Reactivity with other species has not been tested.

Physical form

Serum with 0.05% NaN3

Preparation Note

Maintain at -20°C in undiluted aliquots for up to 1 year after date of receipt.

Analysis Note

Control
C2C12 Tissue lysate
Routinely tested on C2C12 tissue lysate
Lot Specific Tested Application 1: Western Blot

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library