Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
9
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
rat, mouse
technique(s)
immunohistochemistry: suitable (paraffin), western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
mouse ... Vegfa(22339)
rat ... Vegfa(83785)
General description
The human VEGFA, a 45 kDa homodimeric glycoprotein, is one of the most potent angiogenic factors involved in endothelial cell proliferation, migration, and secretion of MMPs. It also serves as an important survival factor for endothelial cells, inhibiting apoptosis. VEGFA belongs to a family that includes VEGFA, VEGFB, VEGFC, VEGFD, and placental growth factor (PLGF). In humans four prominent isoforms of VEGFA are recognized, VEGF121, VEGF165, VEGF189, and VEGF206. In addition, two other less frequent splice variants, VEGF145 and VEGF183, have also been reported. The properties of native VEGF are very similar to VEGF165. Of all the different isoforms of VEGF, the VEGF165 has the highest bioavailability and biological potency. VEGFA action is mediated by VEGF receptor-1 (VEGFR-1) and VEGFR-2. VEGFR-1 (180 kDa, Flt-1) expression is upregulated by hypoxic conditions in the inner core of tumor mass. VEGFR-2 (200 kDa, KDR) is a major mediator of mitogenic and angiogenic permeability enhancing effects of VEGF. It is also reported to mediate survival signals in endothelial cells.
~27 kDa observed
Immunogen
Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to mouse VEGF-A near the N-terminus.
Application
Anti-VEGF-A Antibody is an antibody against VEGF-A for use in Western Blotting and Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected VEGF-A in mouse kidney, mouse cerebellum, mouse spleen, and rat kidney tissue.
Research Category
Signaling
Signaling
Research Sub Category
Metabolic Hormones & Receptors
Metabolic Hormones & Receptors
Physical form
Affinity
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blotting in mouse lung tissue lysate.
Western Blotting Analysis: 2.0 µg/mL of this antibody detected VEGF-A in 10 µg of mouse lung tissue lysate.
Western Blotting Analysis: 2.0 µg/mL of this antibody detected VEGF-A in 10 µg of mouse lung tissue lysate.
Other Notes
Concentration: Please refer to lot specific datasheet.
Replaces: AB1876
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.