Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.22
UNSPSC Code:
12352209
Pricing and availability is not currently available.
Quality Segment
product line
NovaSyn® TG, Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
15-25°C
General description
Pre-loaded resin for synthesis of protected peptide acids containing a C-terminal glutamic acid amino-acid residue by Fmoc SPPS. Cleavage of the protected fragment is effected by treatment with 20% TFE in DCM. The base NovaSyn TG is a composite of low cross-linked polystyrene and 3000-4000 M.W. polyethylene glycol. Peptide synthesis occurs at the ends of the PEG chains which have been functionalized with a TFA-labile Wang-type linker. NovaSyn TG resins are ideal for the synthesis of difficult and long peptides as the PEG helps solvate the growing peptide chains and increase site isolation, thereby improving synthesis efficiency and peptide product quality. The 90 μm beads have a narrow size distribution, high diffusion rates and excellent swelling properties across a wide range of solvents from toluene to water, making them ideally suited to the synthesis of serial and parallel peptide
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Analysis Note
Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.10 - 0.30 mmol/g
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
The base resin is TentaGel (90 µm), functionalized with carboxy- tritylchloride.
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.10 - 0.30 mmol/g
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
The base resin is TentaGel (90 µm), functionalized with carboxy- tritylchloride.
Other Notes
Replaces: 04-12-2710
Legal Information
NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
1 of 1
This Item | |||
|---|---|---|---|
| description Novabiochem® | description Novabiochem® | description Novabiochem® | description Novabiochem® |
| form beads | form beads | form beads | form solid |
| functional group Fmoc | functional group Fmoc | functional group Fmoc | functional group Fmoc |
| product line NovaSyn® TG, Novabiochem® | product line NovaSyn® TG, Novabiochem® | product line NovaSyn® TG, Novabiochem® | product line NovaSyn® TG, Novabiochem® |
| storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 2-8°C |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| application(s) peptide synthesis | application(s) peptide synthesis | application(s) peptide synthesis | application(s) peptide synthesis |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



