8.55101
REM resin (50-100 mesh)
Novabiochem®
Synonym(s):
Acryloyl Wang resin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
13111023
NACRES:
NA.22
Recommended Products
Quality Level
product line
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
reactivity: amine reactive
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
storage temp.
2-30°C
General description
Novabiochem™′s 50-100 mesh REM resin [1] is an exceptionally high quality resin, designed specially for use in IRORI Kans and other parallel synthesis applications that require supports with reproducible substitution and swelling properties containing minimal leachable impurities. This material is derived from poly(chloromethylstyrene-styrene-divinylbenzene) manufactured to ISO 9001 standards. Unlike other Merrifield supports, which are prepared by chloromethylation of polystyrene, this matrix is prepared by co-polymerization of chloromethylstyrene, styrene and divinylbenzene. Since the amount of chloromethylstyrene added to the polymerization reaction can be precisely controlled, this leads to a highly defined resin with little batch to batch variation in functionality. The swelling characteristics of such resins are also more reproducible since the process of manufacture does not introduce additional cross-linking, as is the case with resins produced by chloromethylation. For applications of REM resin in SPOS see references [2-4].
Literature references
[1] A. Brown, et al. (1997) J. Am. Chem. Soc., 119, 3288.
[2] J. Cottney, et al. (1999) Bioorg. Med. Chem. Lett., 9, 1323.
[3] A. Born, et al. (2001) Bioorg. Med. Chem. Lett., 11, 2351.
[4] S. Caix-Haumesser, et al. (2001) Tetrahedron Lett., 42, 3721.
Literature references
[1] A. Brown, et al. (1997) J. Am. Chem. Soc., 119, 3288.
[2] J. Cottney, et al. (1999) Bioorg. Med. Chem. Lett., 9, 1323.
[3] A. Born, et al. (2001) Bioorg. Med. Chem. Lett., 11, 2351.
[4] S. Caix-Haumesser, et al. (2001) Tetrahedron Lett., 42, 3721.
Linkage
Replaces: 01-64-0393
Analysis Note
Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined by elemental analysis of chlorine after derivatization with 4-chloro-benzylamine): 0.80 - 1.20 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene-1 % DVB), 50 - 100 mesh.
Appearance of substance (visual): beads
Loading (determined by elemental analysis of chlorine after derivatization with 4-chloro-benzylamine): 0.80 - 1.20 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene-1 % DVB), 50 - 100 mesh.
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service