Skip to Content
Merck

8.55008

Sieber Amide resin

Novabiochem®

Synonym(s):

Sieber Amide resin, 9-Fmoc-amino-xanthen-3-yloxy-Merrifield resin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
NACRES:
NA.22
UNSPSC Code:
12352111
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

A hyper acid-labile resin for the Fmoc SPPS of protected peptide amides via mild 1% TFA cleavage [1,2]. The resin can be readily reductively alkylated to provide a support suited to the synthesis of secondary carboxamides [3].This resin has also been employed to produce protected peptide fragments in which the C-terminal carboxylic acid group is blocked as a hydroxymethylphenoxy-β-alaninamide ester [4] .

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Application

  • Determining the Hydrophobicity Index of Protected Amino Acids and Common Protecting Groups: Discusses the synthesis of peptide sequences using Sieber Amide resin for its acid-labile properties. (Gavva et al., 2023).
  • Smart biomaterials: Surfaces functionalized with proteolytically stable osteoblast-adhesive peptides: Utilizes Sieber Amide resin for surface functionalization techniques in biomaterials. (Zamuner et al., 2017).
  • A 99mTc-tricine-HYNIC-labeled peptide targeting the melanocortin-1 receptor for melanoma imaging: Describes the use of Sieber Amide resin in the synthesis of radiolabeled peptides for diagnostic imaging. (Shamshirian et al., 2016).

Analysis Note

Color (visual): white to light yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 0.80 mmol/g
Swelling Volume (in DMF): lot specific result
Swelling Volume (in CH₂Cl₂): lot specific result
Total swelling volume acc. Houben Weyl (in CH2Cl2): ≥ 5.2 ml/g
Total swelling volume acc. Houben Weyl (in DMF): ≥ 5.2 ml/g
The polymer matrix is copoly (styrene - 1% DVB) 100 -200 mesh

Other Notes

Replaces: 01-64-0059

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
855008000104027269109448
855008002504027269109462
855008000504027269109455
855008010004027269182380
855008000004027269109431