Skip to Content
Merck

Skip To

8.55003

Rink Amide MBHA resin (100-200 mesh)

Novabiochem®

Synonym(s):

Rink Amide MBHA resin (100-200 mesh), 4-(2′,4′-Dimethoxyphenyl-Fmoc-aminomethyl)-phenoxyacetamido-norleucyl-MBHA resin (100-200 mesh)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.22
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-30°C

SMILES string

N(C(c7c(cc(cc7)OC)OC)c4ccc(cc4)OCC(=O)NC(c6ccc(cc6)C)c5ccc(cc5)C)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C47H44N2O6/c1-30-13-17-32(18-14-30)45(33-19-15-31(2)16-20-33)48-44(50)29-54-35-23-21-34(22-24-35)46(41-26-25-36(52-3)27-43(41)53-4)49-47(51)55-28-42-39-11-7-5-9-37(39)38-10-6-8-12-40(38)42/h5-27,42,45-46H,28-29H2,1-4H3,(H,48,50)(H,49,51)

InChI key

JDDWRLPTKIOUOF-UHFFFAOYSA-N

General description

This support comprises the modified Rink amide linker attached via norleucine to MBHA resin, and is an ideal tool for the Fmoc SPPS of peptide amides. Cleavage from this resin can be effected by a single step treatment with 95% TFA, providing peptide amides in high yields and purities.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Application


  • Liquid chromatography-tandem mass spectrometry for forensic analysis: LSD-D₃ is utilized as an internal standard in the liquid chromatography-tandem mass spectrometry determination of LSD and its metabolites in forensic samples, enhancing accuracy and precision in the analytical results (Johansen and Jensen, 2005).

  • Isotope dilution mass spectrometry in urine testing: Employed in the high-performance liquid chromatography-isotope dilution mass spectrometry (IDMS) method, LSD-D₃ serves as a crucial internal standard for the quantification of lysergide (LSD) in urine, aiding in precise and accurate drug testing (White et al., 1999).

  • Automated extraction and LC/MS/MS confirmation: LSD-D₃ is used in the automated extraction of lysergic acid diethylamide from various biological matrices, followed by confirmation using LC/MS/MS. This application highlights its role in improving the efficiency and reliability of toxicological analyses (de Kanel et al., 1998).

Analysis Note

Colour (visual): white to light yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 0.90 mmol/g
TNBS-Test: passes test
Swelling Volume (in DMF): lot specific result
Swelling Volume (in CH₂Cl₂): lot specific result
Total swelling volume acc. Houben Weyl (in CH2Cl2): ≥ 5.2 ml/g
Total swelling volume acc. Houben Weyl (in DMF): ≥ 5.2 ml/g
The polymer matrix is copoly (styrene-1 % DVB), 100 -200 mesh.

Other Notes

Replaces: 01-64-0037

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
855004855130855001
description

Novabiochem®

description

Novabiochem®

description

Novabiochem®

description

Novabiochem®

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

functional group

amine

functional group

amine

functional group

amine

functional group

amine

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

Quality Level

400

Quality Level

400

Quality Level

400

Quality Level

400

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–4 of 4 Questions  
  1. How can I determine the exact loading capacity of the MBHA Rink Amide resin, as a range for the loading capacity is given in the specification sheet instead of a one?

    1 answer
    1. The exact loading is present on the batch specific certificate of analysis (COA). The specification sheet only shows the acceptance range for each batch, while the COA will show the testing results for a given batch.

      Helpful?

  2. Could you please clarify what is meant by "100-200 mesh"? Does it indicate that the substitution degree of the resin is between 0.1 mmol/g and 0.2 mmol/g?

    1 answer
    1. he term "100-200 mesh" for Rink Amide MBHA resin refers to the particle size of the resin beads, not the substitution degree. The substitution degree for this resin is specified separately and is typically in the range of 0.40–0.90 mmol/g. Therefore, "100-200 mesh" does not indicate a substitution degree of 0.1–0.2 mmol/g.

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  4. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters