Skip to Content

8.53064

Boc-γ-Abu-OH

Novabiochem®

Synonym(s):

Boc-γ-Abu-OH, t.-Boc-4-aminobutanoic acid,t.-Boc-GABA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H17NO4
CAS Number:
Molecular Weight:
203.24
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-30°C

SMILES string

N(CCCC(=O)O)C(=O)OC(C)(C)C

InChI

1S/C9H17NO4/c1-9(2,3)14-8(13)10-6-4-5-7(11)12/h4-6H2,1-3H3,(H,10,13)(H,11,12)

InChI key

HIDJWBGOQFTDLU-UHFFFAOYSA-N

General description

Standard building block for introduction of γ-aminobutyric acid amino-acid residues by Boc SPPS.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Water (K. F.): ≤ 1.5 %
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-0116

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library