Skip to Content
Merck

8.52124

Fmoc-Lys(Alloc)-OH

≥98.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Lys(Alloc)-OH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H28N2O6
CAS Number:
Molecular Weight:
452.50
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fmoc-Lys(Alloc)-OH, Novabiochem®

Quality Segment

product line

Novabiochem®

assay

≥96.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

87-91 °C

application(s)

peptide synthesis

functional group

amine

storage temp.

2-30°C

SMILES string

N([C@@H](CCCCNC(=O)OCC=C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C25H28N2O6/c1-2-15-32-24(30)26-14-8-7-13-22(23(28)29)27-25(31)33-16-21-19-11-5-3-9-17(19)18-10-4-6-12-20(18)21/h2-6,9-12,21-22H,1,7-8,13-16H2,(H,26,30)(H,27,31)(H,28,29)/t22-/m0/s1

InChI key

OJBNDXHENJDCBA-QFIPXVFZSA-N

General description

Orthogonally-protected building block for the synthesis of branched and cyclic peptides and peptides modified at the Lys side chain by Fmoc SPPS. The Alloc group can be selectively removed in the presence of standard Fmoc- and t-butyl-based protecting groups by treatment with Pd(Ph3P)4/CHCl3/AcOH/NMM.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Guide to Selection of Orthogonally -Protected Building Blocks for Fmoc SPPS

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Optical rotation α 25/D (c=1 in DMF): -13.5 - -11.0 °
Purity (TLC(157A)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 96.0 %
Water (K. F.): ≤ 1.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-1305

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Fmoc-Lys(Alloc)-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library