Skip to Content
Merck

8.52033

Fmoc-Hyp-OH

Novabiochem®

Synonym(s):

Fmoc-Hyp-OH, N-α-Fmoc-L-trans-4-hydroxyproline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H19NO5
CAS Number:
Molecular Weight:
353.37
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥95.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

189 °C

application(s)

peptide synthesis

functional group

hydroxyl

storage temp.

2-30°C

SMILES string

N4([C@@H](C[C@H](C4)O)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C20H19NO5/c22-12-9-18(19(23)24)21(10-12)20(25)26-11-17-15-7-3-1-5-13(15)14-6-2-4-8-16(14)17/h1-8,12,17-18,22H,9-11H2,(H,23,24)/t12-,18+/m1/s1

InChI key

GOUUPUICWUFXPM-XIKOKIGWSA-N

General description

Building block for introduction of hydroxyproline amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Color (visual): white to yellow to brown
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 95.0 %
Water (K. F.): ≤ 1.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-1068

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Fmoc-Hyp-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library