Skip to Content
Merck

Skip To

8.52017

Fmoc-Pro-OH

Novabiochem®

Synonym(s):

Fmoc-Pro-OH, N-α-Fmoc-L-proline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C20H19NO4
CAS Number:
Molecular Weight:
337.37
UNSPSC Code:
12352209
EC Index Number:
276-259-0
NACRES:
NA.22
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥94.0% (acidimetric), ≥98% (TLC), ≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

112-115 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-30°C

SMILES string

N4(C(CCC4)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C20H19NO4/c22-19(23)18-10-5-11-21(18)20(24)25-12-17-15-8-3-1-6-13(15)14-7-2-4-9-16(14)17/h1-4,6-9,17-18H,5,10-12H2,(H,22,23)

InChI key

ZPGDWQNBZYOZTI-UHFFFAOYSA-N

General description

High purity Fmoc-protected amino acid for research and process production of peptides, with very low levels of dipeptide, free-amino acids and acetic acid impurities.
Standard building block for introduction of proline amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Fmoc-amino acids for Peptide Production
Cleavage and Deprotection Protocols for Fmoc SPPS

Application


Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Colour index (0,5 M in DMF): ≤ 150 Hazen
Identity (IR): passes test
Enantiomeric purity: ≥ 99.8 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Fmoc-ß-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-β-Ala-Pro-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Pro-Pro-OH (HPLC): ≤ 0.1 % (a/a)
Assay free amino acid (GC): ≤ 0.2 %
Purity (TLC(011A)): ≥ 98 %
Solubility (25 mmole in 50 ml DMF): clearly soluble
Assay (acidimetric): ≥ 94.0 %
Water (K. F.): ≤ 6.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-1031

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
852149852208852021
description

Novabiochem®

description

Novabiochem®

description

Novabiochem®

description

Novabiochem®

form

powder

form

powder

form

powder

form

powder

assay

≥94.0% (acidimetric), ≥99.0% (HPLC), ≥98% (TLC)

assay

≥93.0% (acidimetric), ≥98% (TLC), ≥99.0% (HPLC)

assay

≥85.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

assay

≥98% (TLC), ≥98.0% (acidimetric), ≥99.0% (HPLC)

functional group

Fmoc

functional group

Fmoc

functional group

Fmoc

functional group

Fmoc

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

15-25°C

storage temp.

2-30°C

mp

112-115 °C

mp

-

mp

-

mp

140-145 °C


Still not finding the right product?

Explore all of our products under Fmoc-Pro-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters