Skip to Content
Merck

Skip To

8.22293

2-Nitrobenzaldehyde

for synthesis

Synonym(s):

2-Nitrobenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
2-(NO2)C6H4CHO
CAS Number:
Molecular Weight:
151.12
UNSPSC Code:
12352114
EC Index Number:
209-025-3
NACRES:
NA.22
MDL number:
Assay:
≥99.0% (GC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0% (GC)

autoignition temp.

200 °C (Lit.)

bp

156 °C/20 hPa

mp

43.5 °C

bulk density

350 kg/m3

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1c(cccc1)C=O

InChI

1S/C7H5NO3/c9-5-6-3-1-2-4-7(6)8(10)11/h1-5H

InChI key

CMWKITSNTDAEDT-UHFFFAOYSA-N

Application

2-Nitrobenzaldehyde for synthesis. CAS No. 552-89-6, EC Number 209-025-3.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 41 °C
Melting range (upper value): ≤ 43 °C
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under 2-Nitrobenzaldehyde


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

flash_point_f

230.0 °F

flash_point_c

110 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library