Skip to Content
Merck
All Photos(1)

Key Documents

8.22287

Sigma-Aldrich

Hydrogen peroxide solution

30%, (stabilized) for synthesis

Synonym(s):

Hydrogen peroxide solution, Perhydrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

Hydrogen peroxide 30%, (stabilized) for synthesis

vapor pressure

18 hPa ( 20 °C)
23.3 mmHg ( 30 °C)

Quality Level

form

liquid

reaction suitability

reagent type: oxidant

pH

2-4 (20 °C in H2O)

bp

107 °C/1013 hPa

mp

-26 °C

density

1.11 g/cm3 at 20 °C

storage temp.

2-25°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydrogen peroxide 30% can be used as an oxidizing agent:
  • To synthesize 5-bromo-2-nitropyridine from 5-bromo-2-aminopyridine in the presence of H2SO4.[1]
  • For the oxidation of aromatic and aliphatic sulfides to sulfoxides or sulfones.[2]
  • For the epoxidation of terminal olefins using a mixture of Na2WO4 dihydrate, (aminomethyl)phosphonic acid, and methyltrin-octylammonium hydrogensulfate.[3]
  • To prepare adipic acid via oxidation of cyclohexanone and cyclohexanol in the presence of H2WO4 as a catalyst.[4]
  • To convert cyclopentanone to δ-valerolactone via Baeyer−Villiger-oxidation in the presence of a solid acid catalyst.[5]

Analysis Note

Assay (manganometric): 29.0 - 31.0 %
Density (d 20/4): 1.11 - 1.12 g/cm³

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

not combustible

Flash Point(C)

not combustible


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handling hydrogen peroxide oxidations on a Large Scale: synthesis of 5-bromo-2-nitropyridine
Agosti A, et al.
Organic Process Research & Development, 21(3), 451-459 (2017)
Oxidation of sulfides to sulfoxides and sulfones with 30% hydrogen peroxide under organic solvent-and halogen-free conditions
Sato K, et al.
Tetrahedron, 57(13), 2469-2476 (2001)
A green method of adipic acid synthesis: organic solvent-and halide-free oxidation of cycloalkanones with 30% hydrogen peroxide
Usui Y and Sato K
Green Chemistry, 5(4), 373-375 (2003)
Baeyer-Villiger-oxidation of cyclopentanone with aqueous hydrogen peroxide by acid heterogeneous catalysis
Fischer J and Holderich WF
Applied Catalysis A: General, 180(1-2), 435-443 (1999)
A Practical Method for Epoxidation of Terminal Olefins with 30 Hydrogen Peroxide under Halide-Free Conditions.
Kazuhiko Sato et al.
The Journal of organic chemistry, 61(23), 8310-8311 (1996-11-15)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service