Skip to Content
Merck

8.20643

1-Hexyne

for synthesis

Synonym(s):

1-Hexyne

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H10
CAS Number:
Molecular Weight:
82.14
UNSPSC Code:
12352003
EC Index Number:
211-736-9
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

3535 kPa

Quality Segment

assay

≥98.0% (GC)

form

liquid

autoignition temp.

263 °C

bp

71-72 °C/1013 hPa

mp

-132 °C

transition temp

flash point -20 °C

solubility

0.36 g/L (experimental)

density

0.72 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

C(CC)CC#C

InChI

1S/C6H10/c1-3-5-6-4-2/h1H,4-6H2,2H3

InChI key

CGHIBGNXEGJPQZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.715 - 0.720
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under 1-Hexyne


signalword

Danger

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

-4.0 °F - closed cup

flash_point_c

-20 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library