Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
MDL number:
UNSPSC Code:
12352115
EC Index Number:
202-494-5
NACRES:
NA.22
Form:
powder
vapor pressure
<0.01 Pa ( 20 °C)
Quality Level
form
powder
potency
>16000 mg/kg LD50, oral (Rat)
pH
4-6 (20 °C, 50 g/L in H2O)
bp
188 °C (decomposition,OECD Test Guideline 103)
mp
96.5 °C (Monomer)
density
1.52 g/cm3 at 20 °C (OECD Test Guideline 109)
storage temp.
2-8°C
SMILES string
OCC(=O)CO
InChI
1S/C3H6O3/c4-1-3(6)2-5/h4-5H,1-2H2
InChI key
RXKJFZQQPQGTFL-UHFFFAOYSA-N
Application
- Biosynthesis of catalysts for glycerol oxidation: A study demonstrates the biosynthesis of Au/CuO catalysts using Syringa oblata Lindl. leaf extract for efficient selective oxidation of glycerol to 1,3-dihydroxyacetone, highlighting a sustainable approach in catalyst production (Yuan et al., 2023).
- Photocatalytic hydrogen production: Research on biochars from olive stones as carbonaceous supports in Pt/TiO(2)-carbon photocatalysts explores their application in hydrogen production from aqueous glycerol photoreforming, offering insights into renewable energy sources and the role of dihydroxyacetone in sustainable technologies (Escamilla-Mejía et al., 2023).
Analysis Note
Appearance: white to almost white finecrystalline-freeflowing powder, eventually with granular parts.
Assay (periodatometric): ≥ 98 %
Identity (IR-spectrum): confirms
Assay (periodatometric): ≥ 98 %
Identity (IR-spectrum): confirms
Still not finding the right product?
Explore all of our products under Dihydroxyacetone
Storage Class
13 - Non Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8204820250 | 04022536459981 |
| 8204829999 | 04022536459998 |
| 8204820025 | 04027269912925 |
| 8204826000 | 04054839232312 |
| 8204820100 | 04022536459974 |
| 8204820000 | 04022536459967 |